Skip to Content
Merck
All Photos(1)

Key Documents

48500-U

Supelco

Acenaphthene

analytical standard

Synonym(s):

1,8-Ethylenenaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10
CAS Number:
Molecular Weight:
154.21
Beilstein:
386081
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.32 (vs air)

vapor pressure

10 mmHg ( 131 °C)

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

279 °C (lit.)

mp

90-94 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

C1Cc2cccc3cccc1c23

InChI

1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2

InChI key

CWRYPZZKDGJXCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acenaphthene belongs to the group of polycyclic aromatic hydrocarbons (PAHs), which are considered to be potential environmental pollutants, marine contaminants and possible human carcinogens. PAHs are released during the incomplete combustion of fossil fuels, petroleum production, oil spills and some industrial processes.

Application

Acenaphthene may be used as an analytical reference standard for the determination of the analyte in wastewater, soil samples, aviation jet fuel and air by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of polycyclic aromatic hydrocarbons in waste water by off-line coupling of solid-phase microextraction with column liquid chromatography
Popp P, et al.
Journal of Chromatography A, 897(1-2), 153-159 (2000)
Rapid determination of PAHs in soil samples by HPLC with fluorimetric detection following sonication extraction
Kayali-Sayadi MN, et al.
Fresenius Journal of Analytical Chemistry, 368(7), 697-701 (2000)
Determination of total and polycyclic aromatic hydrocarbons in aviation jet fuel
Bernabei M, et al.
Journal of Chromatography A, 985(1-2), 197-203 (2003)
G Bieniek
Journal of chromatography. A, 891(2), 361-365 (2000-10-24)
An attempt was made to establish a method for the simultaneous determination of ethylbenzene, indan, indene and acenaphthene by capillary gas chromatography with flame ionization detection. The air was sampled on charcoal tubes and extracted with carbon disulfide-methanol (60:1, v/v).
Zhipei Qin et al.
Journal of chromatography. A, 1196-1197, 89-95 (2008-04-22)
The characterization of a poly(dimethylsiloxane) (PDMS) thin film for active extraction of some polycyclic aromatic hydrocarbons was investigated in both a 1-L aqueous solution and a flow-through system. The thin film was attached to an electric bench-drill at a constant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service