Skip to Content
Merck
All Photos(1)

Key Documents

46959-U

Supelco

Retinol palmitate

analytical standard

Synonym(s):

Vitamin A palmitate, all trans-Retinol palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60O2
CAS Number:
Molecular Weight:
524.86
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

feature

standard type fat soluble vitamin

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-8°C

InChI

1S/C36H60O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-25-35(37)38-30-28-32(3)23-20-22-31(2)26-27-34-33(4)24-21-29-36(34,5)6/h20,22-23,26-28H,7-19,21,24-25,29-30H2,1-6H3/b23-20+,27-26+,31-22+,32-28+

InChI key

VYGQUTWHTHXGQB-FFHKNEKCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Retinol palmitate is suitable for use in the retention identification of the analyte when using HPLC and GC. Not intended for use as an activity reference standard. It has been thoroughly evaluated to ensure the utmost quality. Neat, unless otherwise noted. Certificate of Compostion provided with the purchase.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M M Ramalho et al.
Journal of dairy science, 95(10), 5476-5484 (2012-08-21)
In Portugal, commercial milk is obtained almost entirely from cows of the Holstein breed. However, other breeds may also show dairy aptitude, and produce milk of good quality. The aim of this study was to investigate the effect of the
Anta Agne-Djigo et al.
The Journal of nutrition, 142(11), 1991-1996 (2012-09-28)
To alleviate vitamin A (VA) deficiency (VAD) in Senegal, understanding the relationship between VA status of lactating women and their 6-mo-old infants is important. This study measured 6-mo-old infants' VA intake from human milk and assessed the VA status of
Mirela Donato Gianeti et al.
Molecules (Basel, Switzerland), 17(2), 2219-2230 (2012-02-24)
Chemically stable ester derivatives of vitamins A, C and E have become a focus of interest for their role in the satisfactory results in skin aging treatments. Accordingly, the aim of this study was to evaluate the physical and chemical
Eliot L Berson et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 130(6), 707-711 (2012-02-15)
To evaluate whether a diet high in long chain ω-3 fatty acids can slow the rate of visual acuity loss among patients with retinitis pigmentosa receiving vitamin A palmitate. We calculated dietary intake from questionnaires completed annually by 357 adult
Zhu Kai et al.
Journal of nanoscience and nanotechnology, 11(9), 7593-7602 (2011-11-22)
Candida sp.99-125 lipase was suitable for transesterification of fats and oils to produce fatty acid methyl ester. The adsorption of Candida sp.99-125 lipase onto the fiber-like SBA-15 mesoporous material has been studied. The unaltered structural order of the fiber-like SBA-15

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service