Skip to Content
Merck
All Photos(1)

Key Documents

442475

Supelco

Benzo[e]pyrene

analytical standard

Synonym(s):

1,2-Benzpyrene, 4,5-Benzpyrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1911334
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

177-180 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

c1ccc2c(c1)c3cccc4ccc5cccc2c5c34

InChI

1S/C20H12/c1-2-8-16-15(7-1)17-9-3-5-13-11-12-14-6-4-10-18(16)20(14)19(13)17/h1-12H

InChI key

TXVHTIQJNYSSKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzo[e]pyrene is a pentacyclic aromatic hydrocarbon contaminant. It is a structural isomer of the potent carcinogen benzo[a]pyrene.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of sulfate and glucoside conjugates of benzo [e] pyrene by Cunninghamella elegans
Pothuluri.V.J, et al.
Applied Microbiology and Biotechnology, 45, 677-683 (1996)
Photochemical transformations of benzo [e] pyrene in solution and adsorbed on silica gel and alumina surfaces
Fioressi S and Arce R
Environmental Science & Technology, 39, 3646-3655 (2005)
Silvina Fioressi et al.
Environmental science & technology, 39(10), 3646-3655 (2005-06-16)
The photodegradation of benzo[e]pyrene (BeP), a ubiquitous polycyclic aromatic hydrocarbon (PAH) contaminant, was investigated in solution and adsorbed on surfaces modeling the atmospheric particulate matter to provide fundamental information that could help to clarify its fate in the atmosphere. Diones
Joris J H Haftka et al.
Environmental toxicology and chemistry, 27(7), 1526-1532 (2008-02-12)
The uptake kinetics of fluorene, phenanthrene, fluoranthene, pyrene, and benzo[e]pyrene by solid-phase microextraction fibers was studied in the presence of dissolved organic matter (DOM) obtained from sediment pore water and resulted in increased fiber absorption and desorption rate coefficients. Compared
Lawrence R Curtis et al.
Toxicology and applied pharmacology, 254(1), 1-7 (2011-05-10)
This study assessed the role of aryl hydrocarbon receptor (AHR) affinity, and cytochrome P4501A (CYP1A) protein and activity in polyaromatic hydrocarbon (PAH)-induced oxidative stress. In the 1-100nM concentration range benzo[a]pyrene (BaP) but not benzo[e]pyrene (BeP) competitively displaced 2nM [(3)H]2, 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service