Skip to Content
Merck
All Photos(1)

Key Documents

442404

Supelco

4-Bromofluorobenzene

analytical standard

Synonym(s):

1-Bromo-4-fluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4F
CAS Number:
Molecular Weight:
175.00
Beilstein:
774102
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.527 (lit.)

bp

150 °C (lit.)

mp

−16 °C (lit.)

density

1.593 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Fc1ccc(Br)cc1

InChI

1S/C6H4BrF/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

AITNMTXHTIIIBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers 4-bromofluorobenzene (1-Bromo-4-fluorobenzene) for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyzes.

Application


4-Bromofluorobenzene can act like a surrogate standard for the:
  • determination of toxicity of diesel and water accommodated diesel fraction (WAF), towards microalgal species like Pseudokirchneriella subcapitata and Chlorella sp.MM3, using gas chromatography coupled with mass spectrometry (GC-MS).
  • determination of the chemicals like benzene, toluene, ethylbenzene and xylene (BTEX) and their stable carbon isotopic values, in gasoline contaminated ground-water, using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concentrations and stable isotope values of BTEX in gasoline-contaminated groundwater
Kelley.AC, et al.
Environmental Science & Technology, 31, 2469-2472 (1997)
Toxicity of diesel water accommodated fraction toward microalgae, Pseudokirchneriella subcapitata and Chlorella sp. MM3
Ramadass K, et al.
Ecotox. Environ. Safety, 142, 538-543 (2017)
Marcelo Volpon Santos et al.
Developmental neuroscience, 41(1-2), 34-43 (2019-04-19)
Hydrocephalus is a multifactorial disease, affecting the dynamics of cerebrospinal fluid (CSF) and leading to severe neurological impairment in children; in spite of the recent advances in hydrocephalus research, it has many physiopathological aspects that still remain poorly understood, especially
Qian Luo et al.
Journal of chromatography. A, 1365, 45-53 (2014-09-27)
When iodide and natural organic matter are present in raw water, the formation of iodo-trihalomethanes (Iodo-THMs), haloacetonitriles (HANs), and halonitromethanes (HNMs) pose a potential health risk because they have been reported to be more toxic than their brominated or chlorinated
Michael Phillips et al.
Health physics, 108(5), 538-546 (2015-03-27)
There is widespread interest in the development of tools to estimate radiation exposures. Exhaled breath provides a novel matrix for assessing biomarkers that could be correlated with exposures. The use of exhaled breath for estimating radiation exposure is warranted, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service