B60428
1-Bromo-4-chlorobenzene
99%
Synonym(s):
1-Chloro-4-bromobenzene, 4-Bromo-1-chlorobenzene, 4-Bromochlorobenzene, 4-Bromophenyl chloride, p-Chlorophenyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
BrC6H4Cl
CAS Number:
Molecular Weight:
191.45
Beilstein:
1099014
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
crystals
bp
196 °C (lit.)
mp
64-67 °C (lit.)
SMILES string
Clc1ccc(Br)cc1
InChI
1S/C6H4BrCl/c7-5-1-3-6(8)4-2-5/h1-4H
InChI key
NHDODQWIKUYWMW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shigeo Hayashi et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 846-867 (2014-02-13)
Because of the pivotal role of cyclooxygenase (COX) in the inflammatory processes, non-steroidal anti-inflammatory drugs (NSAIDs) that suppress COX activities have been used clinically for the treatment of inflammatory diseases/syndromes; however, traditional NSAIDs exhibit serious side-effects such as gastrointestinal damage
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service