Z742699
SureTemp® Dual Convection Incubator stainless steel shelf
stainless steel shelf for 130 L incubator
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NB.77
Recommended Products
material
stainless steel
manufacturer/tradename
Benchmark Scientific H2505-130SH
inside W × D × H
55 cm × 55 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Extra stainless steel shelf for Benchmark SureTemp 130L.
Legal Information
SureTemp is a registered trademark of Benchmark Scientific, Inc.
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service