Z740235
TLC Plastic Sheets
L × W 20 cm × 20 cm, silica gel 60 matrix, binder, Organic Polymer, fluorescent indicator
Synonym(s):
Plastic-based TLC plates
About This Item
Recommended Products
material
plastic support
silica gel 60 matrix
Quality Level
feature
binder Organic Polymer
fluorescent indicator
packaging
pkg of 25 ea
manufacturer/tradename
EMD Millipore® 1.05735.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
200 μm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | RAB0213 | RAB0149 | RAB0182 |
---|---|---|---|
human | human | human | human |
sample type urine | sample type tissue lysate | sample type cell culture supernatant(s) | sample type cell culture supernatant(s) |
inter-assay cv: <12% | inter-assay cv: <12% | inter-assay cv: <12% | inter-assay cv: <12% |
colorimetric | colorimetric | colorimetric | colorimetric |
ELISA: suitable, capture ELISA: suitable | ELISA: suitable, capture ELISA: suitable | ELISA: suitable, capture ELISA: suitable | ELISA: suitable, capture ELISA: suitable |
Application
- [Development of salicylic acid dipstick test]: Utilizing TLC plastic sheets, this research contributes to the field of clinical diagnostics by developing a quick and effective dipstick test for salicylic acid, facilitating rapid medical assessments (Kirihara, Ishizawa, Ishiwata, 2005).
- Amino acid sequence of an excitatory insect-selective toxin from venom of the scorpion Buthus martensi Karsch: This study employs TLC plastic sheets for the separation of complex protein mixtures, enhancing our understanding of bioactive compounds in venom research, which is critical for developing new pharmaceuticals (Ji, Mansuelle, Xu, Granier, Kopeyan, Terakawa, Rochat, 1994).
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service