Z265500
Analtech TLC Uniplates™: silica gel matrix
L × W 20 cm × 20 cm, binder, Organic Polymer, fluorescent indicator
Synonym(s):
Silica Gel TLC Plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
glass support
silica gel 60 matrix
Quality Level
feature
binder Organic Polymer
fluorescent indicator
packaging
pkg of 25 ea
manufacturer/tradename
Analtech 47011
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
250 μm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
General description
Most rugged silica gel plates available.
Legal Information
Uniplates is a trademark of Analtech, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service