Skip to Content
Merck
All Photos(1)

Key Documents

Z719684

Sorenson low binding standard tips

maximum volume 1000 μL, pack of 1000 ea (Racked), sterile

Synonym(s):

low-binding tip, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

Pricing and availability is not currently available.

material

colorless
polypropylene

sterility

sterile

feature

barrier: no

packaging

pack of 1000 ea (Racked)
case of 5000 ea (Racked)

manufacturer/tradename

Sorenson 10230T

maximum volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
T7665SML1741SML2212
TH1020 ≥95% (HPLC)

SML1741

TH1020

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: soluble 5 mg of Tyrphostin 51 in 0.1 ml of solvent, clear, orange to red

solubility

DMSO: 3 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

color

white to very dark gray

color

-

color

white to beige

color

white to beige

General description

Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.

Legal Information

Sorenson is a trademark of Sorenson BioScience, Inc.

  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service