SML2212
RWJ 67657
≥98% (HPLC)
Synonym(s):
3-Butyn-1-ol, 4-[4-(4-fluorophenyl)-1-(3-phenylpropyl)-5-(4-pyridinyl)-1H-imidazol-2-yl], 4-(4-Fluorophenyl)-2-(4-hydroxy-1-butynyl)-1-(3-Phenylpropyl)-5-(4-Pyridyl)imidazole, 4-[4-(4-Fluorophenyl)-1-(3-phenylpropyl)-5-(4-pyridinyl)-1H-imidazol-2-yl]-3-butyn-1-ol, JNJ 3026582
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
OCCC#CC1=NC(C2=CC=C(F)C=C2)=C(C3=CC=NC=C3)N1CCCC4=CC=CC=C4
InChI
1S/C27H24FN3O/c28-24-13-11-22(12-14-24)26-27(23-15-17-29-18-16-23)31(25(30-26)10-4-5-20-32)19-6-9-21-7-2-1-3-8-21/h1-3,7-8,11-18,32H,5-6,9,19-20H2
InChI key
QSUSKMBNZQHHPA-UHFFFAOYSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service