Skip to Content
Merck
All Photos(1)

Key Documents

Z709026

ART® non-filtered tip

HLT XLP, maximum volume 200 μL, racked, non-sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
200 μL
volume:
200 μL
sterility:
non-sterile
feature:
racked
packaging:
pack of 3840 ea (5 packs of 768)
manufacturer/tradename:
Molecular BioProducts 3542

sterility

non-sterile

feature

racked

packaging

pack of 3840 ea (5 packs of 768)

manufacturer/tradename

Molecular BioProducts 3542

technique(s)

PCR: suitable

maximum volume

200 μL

volume

200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Extended length making the bottom of 12 × 75 tubes and vials easier to reach.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service