Skip to Content
Merck
All Photos(1)

Key Documents

Z359971

Sigma-Aldrich

Pellet pestles

Cordless motor

Synonym(s):

Pellet Grinder, Pestle for Pellets, Pestles for Laboratory Use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
eCl@ss:
32080115
NACRES:
NB.22

manufacturer/tradename

Kimble®

battery power

2 AA (included)

Looking for similar products? Visit Product Comparison Guide

General description

Hand-operated or motor-driven grinders for resuspending pellets or disrupting soft tissue in microcentrifuge tubes. Pestle ends are specially designed to mate with 0.5 mL or 1.5 mL microtubes. Shaft is convenient length and diameter for gentle manual back-and-forth rotation or more vigorous motorized operation.
Z359955 will fit the cordless motor product Z359971 only by using the replacement adapter Z359998 for the motor, which is purchased separately.

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantinos Mavridis et al.
Genes, 9(10) (2018-10-05)
Bioassays and molecular diagnostics are routinely used for the monitoring of malaria vector populations to support insecticide resistance management (IRM), guiding operational decisions on which insecticides ought to be used for effective vector control. Previously developed TaqMan assays were optimised
Mikhail A Tyumentsev et al.
Journal of Alzheimer's disease : JAD, 63(3), 1075-1088 (2018-05-02)
Growing evidence suggests that mitochondrial dysfunction is an early event in sporadic Alzheimer's disease (AD), but the impact of mitochondrial dysfunction on the transition from healthy aging to AD remains elusive. Here we estimated the influence of mitochondrial dysfunction on
Juli Choi et al.
Experimental neurobiology, 28(2), 158-171 (2019-05-30)
Gut microbiota play a role in regulating mental disorders, but the mechanism by which gut microbiota regulate brain function remains unclear. Gram negative and positive gut bacteria release membrane-derived extracellular vesicles (EVs), which function in microbiota-host intercellular communication. In the
Chanyoung Song et al.
International journal of nanomedicine, 12, 7501-7517 (2017-10-27)
In this study, we suggest a designer vaccine adjuvant that can mimic the drainage of pathogens into lymph nodes and activate innate immune response in lymph nodes. By the amination of multivalent carboxyl groups in poly-(γ-glutamic acid) (γ-PGA) nanomicelles, the
Deborah J Gorth et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 35(4), 725-737 (2019-12-05)
The inflammatory cytokine tumor necrosis factor alpha (TNFα) is considered to play a key role in the pathogenesis of intervertebral disc disease. To evaluate the importance of this cytokine we examined the inflammatory environment and spinal phenotype of 9-month-old human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service