Z290645
Vacuum filtration assembly funnel tops
Funnel top, funnel capacity 1000 mL
Select a Size
Select a Size
About This Item
Recommended Products
material
glass housing
feature
graduation
packaging
pkg of 1 ea
funnel capacity
1000 mL
membrane diam.
90 mm
neck diam.
90 mm
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service