Z290440
Sigma-Aldrich® vacuum filtration assembly
for 47 mm filters with glass support, #8 stopper, same as Z290432 but without filtration flask
Synonym(s):
Filtration Assembly, Vacuum Filtration Setup
All Photos(2)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Recommended Products
material
glass housing
glass membrane
feature
graduation
packaging
pkg of 1 ea
neck diam.
41 mm
Related Categories
General description
Same as Z290432 but without filtration flask.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany