Skip to Content
Merck
All Photos(2)

Key Documents

X4126

Sigma-Aldrich

Xylene Cyanol FF

BioReagent, for molecular biology

Synonym(s):

Acid blue 147, Cyanol FF, XC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27N2NaO6S2
CAS Number:
Molecular Weight:
538.61
Colour Index Number:
42135
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

Quality Level

product line

BioReagent

form

powder

technique(s)

nucleic acid detection: suitable

solubility

H2O: complete 1 mg/mL, clear, blue

suitability

suitable for nucleic acid staining

storage temp.

room temp

SMILES string

[Na+].CCNc1ccc(cc1C)C(=C2/C=CC(=N\CC)\C(C)=C2)\c3ccc(cc3S(O)(=O)=O)S([O-])(=O)=O

InChI

1S/C25H28N2O6S2.Na/c1-5-26-22-11-7-18(13-16(22)3)25(19-8-12-23(27-6-2)17(4)14-19)21-10-9-20(34(28,29)30)15-24(21)35(31,32)33;/h7-15,26H,5-6H2,1-4H3,(H,28,29,30)(H,31,32,33);/q;+1/p-1/b25-19-,27-23-;

InChI key

VVLFAAMTGMGYBS-KRQUPCAFSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Xylene cyanol is often used as a tracking dye during agarose or polyacrylamide gel electrophoresis. It has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.

Application

Xylene Cyanol FF is used as a:
  • histological stain and as tracker dye in DNA sequencing and electrophoresis
  • gel-loading dye
  • formamide RNA loading dye
  • tracking dye in native polyacrylamide gel electrophoresis (PAGE) loading buffer

Biochem/physiol Actions

Xylene Cyanol FF acts as nucleic acid stain.

Reconstitution

A 6x Gel Loading Buffer can be made using standard recipes, including 0.05% xylene cyanol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P N Hengen
Trends in biochemical sciences, 20(5), 202-203 (1995-05-01)
Methods and reagents is a unique monthly column that highlights current discussions in the newsgroup bionet.molbio.methods-reagents, available on the Internet. This month's column describes how to make an electrophoresis system from common household items. For details on how to partake
Nicolás Cara et al.
Bio-protocol, 10(13), e3671-e3671 (2021-03-05)
Methylation-Sensitive Amplification Polymorphism (MSAP) is a versatile marker for analyzing DNA methylation patterns in non-model species. The implementation of this technique does not require a reference genome and makes it possible to determine the methylation status of hundreds of anonymous
Design and self-assembly of siRNA-functionalized RNA nanoparticles for use in automated nanomedicine
<BIG>Afonin KA, et al.</BIG>
Nature Protocols, 6, 2022-2022 (2011)
Cell-internalization SELEX: method for identifying cell-internalizing RNA aptamers for delivering siRNAs to target cells
<BIG>Thiel WH, et al.</BIG>
Methods in Molecular Biology, 187-199 (2015)
André Vasconcelos et al.
The breast journal, 8(4), 226-229 (2002-07-09)
Breast cancer is the most frequent neoplasm in women. Expression of the estrogen receptor (ER) has a key role in breast cancer; the ER gene is located at chromosome 6q24-q27 and is made up of 8 exons with a total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service