Skip to Content
Merck
All Photos(2)

Key Documents

V7264

Sigma-Aldrich

Venlafaxine hydrochloride

≥98% (HPLC), powder

Synonym(s):

(+/-)-1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, Effexor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO2 · HCl
CAS Number:
Molecular Weight:
313.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white

solubility

H2O: >10 mg/mL

originator

Wyeth

SMILES string

Cl[H].COc1ccc(cc1)C(CN(C)C)C2(O)CCCCC2

InChI

1S/C17H27NO2.ClH/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14;/h7-10,16,19H,4-6,11-13H2,1-3H3;1H

InChI key

QYRYFNHXARDNFZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Venlafaxine is a dual serotonin-noradrenaline reuptake inhibitor that displays Ki values of 82 and 2480 nM respectively. It is a general class antidepressant.
Venlafaxine is an antidepressant. The mechanism of the antidepresant action of venlafaxine in humans is associated with its potentiation of neurotransmitter activity in the CNS. Venlafaxine is a potent inhibitor of neuronal serotonin and norepinephrine reuptake and weak inhibitor of dopamine reuptake. Venlafaxine has no significant activity for muscarinic, histaminergic, or α-1 adrenergic receptors in vitro. Venlafaxine does not possess MAO inhibitor activity.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Freixa et al.
Frontiers in microbiology, 9, 1426-1426 (2018-07-19)
Organic micro-contaminants (OMCs) enter in freshwaters and interact with other contaminants such as carbon nanoparticles, becoming a problem of unknown consequences for river ecosystems. Carbon nanoparticles (as fullerenes C60) are good adsorbents of organic contaminants and their interaction can potentially
Mara Z Vitolins et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(32), 4092-4098 (2013-10-02)
Hot flashes occur in approximately 80% of androgen-deprived men. Few intervention studies have been conducted to relieve hot flashes in men. Eligible androgen-deprived men were randomly assigned to one of four daily regimens (2 × 2 factorial design) for 12
M Batista et al.
Clinical toxicology (Philadelphia, Pa.), 51(2), 92-95 (2013-01-11)
Venlafaxine is a bicyclic antidepressant that may be associated with severe cardiotoxicity following large overdose. The purpose of this short case series is to present different patterns of venlafaxine-related cardiotoxicity and to discuss the potential mechanisms. Between January 2010 and
Yaal Lester et al.
Water research, 47(13), 4349-4356 (2013-06-15)
Wastewater from a pharmaceutical formulation facility (TevaKS, Israel) was treated with a biological activated-sludge system followed by ozonation. The goal was to reduce the concentrations of the drugs carbamazepine (CBZ) and venlafaxine (VLX) before discharging the wastewater to the municipal
Swati Pund et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(1-2), 195-201 (2012-11-20)
Venlafaxine, a dual acting antidepressant is a new therapeutic option for chronic depression. Depression is a common mental disorder associated with the abnormalities in neuronal transport in the brain. Since the nose-to-brain pathway has been indicated for delivering drugs to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service