Skip to Content
Merck
All Photos(1)

Key Documents

D2069

Sigma-Aldrich

Desvenlafaxine hydrochloride

≥98% (HPLC), powder

Synonym(s):

4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-phenol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NO2 · HCl
CAS Number:
Molecular Weight:
299.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >20 mg/mL
H2O: >20 mg/mL

originator

Wyeth

storage temp.

2-8°C

SMILES string

Cl.CN(C)CC(c1ccc(O)cc1)C2(O)CCCCC2

InChI

1S/C16H25NO2.ClH/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16;/h6-9,15,18-19H,3-5,10-12H2,1-2H3;1H

InChI key

IMWPSXHIEURNKZ-UHFFFAOYSA-N

Application

Desvenlafaxine hydrochloride has been used to evaluate if its occurrence in drinking water at environmental concentrations could induce neuronal gene expression.

Biochem/physiol Actions

Desvenlafaxine hydrochloride is a dual SERT/NET reuptake inhibitor. It is a major active metabolite of venlafaxine (#542); brain penetrant; Ki = 40 nM and 122 nM for hSERT and hNET, respectively (weak binding at the hDAT). Inhibition of [3H]5-HT or [3H]NE uptake for the hSERT or hNET produced IC50 values of 47 and 531 nM.
Desvenlafaxine hydrochloride is a psychoactive drug. Incidence of this drug in environmental samples might induce in vitro gene expression associated with neurological disorders.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dysregulation of autism-associated synaptic proteins by psychoactive pharmaceuticals at environmental concentrations
Kaushik G, et al.
Neuroscience Letters, 661(3), 143-148 (2017)
Psychoactive pharmaceuticals at environmental concentrations induce in vitro gene expression associated with neurological disorders
Kaushik G, et al.
BMC Genomics, 17(3), 435-435 (2016)
Alessandro Atzei et al.
International journal of environmental research and public health, 18(13) (2021-07-03)
Humans are exposed daily to complex mixtures of chemical substances via food intake, inhalation, and dermal contact. Developmental neurotoxicity is an understudied area and entails one of the most complex areas in toxicology. Animal studies for developmental neurotoxicity (DNT) are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service