Skip to Content
Merck
All Photos(1)

Key Documents

UC179

Sigma-Aldrich

(R)-(−)-Nirvanol

Synonym(s):

(R)-(−)-5-Ethyl-5-phenyl-2,4-imidazolidinedione, (R)-(−)-5-Ethyl-5-phenylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

off-white

solubility

DMF: soluble

storage temp.

2-8°C

SMILES string

CC[C@@]1(NC(=O)NC1=O)c2ccccc2

InChI

1S/C11H12N2O2/c1-2-11(8-6-4-3-5-7-8)9(14)12-10(15)13-11/h3-7H,2H2,1H3,(H2,12,13,14,15)/t11-/m1/s1

InChI key

UDTWZFJEMMUFLC-LLVKDONJSA-N

Biochem/physiol Actions

CYP2B6 metabolite of (S)-(+)-mephenytoin; anticonvulsant; hypnotic.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Nirvanol is soluble in DMF.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Inaba et al.
American journal of human genetics, 38(5), 768-772 (1986-05-01)
A genetic polymorphism characterized by deficient drug oxidation exists for the hydroxylation of mephenytoin. This deficiency was first recognized in a family study that suggested an autosomal recessive pattern of inheritance. To confirm the observation, we investigated 28 relatives of
R W Nims et al.
Chemical research in toxicology, 6(2), 188-196 (1993-03-01)
The pharmacodynamics of rat hepatic cytochrome P450 2B (P450 2B) induction by phenobarbital (PB) and two structural congeners, dl-5-ethyl-5-phenylhydantoin (EPH) and dl-5-ethyl-5-phenyloxazolidinedione (EPO), were investigated. The in vivo induction of P450 2B was probed in F344/NCr rats by measuring immunoreactive
J W Ko et al.
Drug metabolism and disposition: the biological fate of chemicals, 26(8), 775-778 (1998-08-11)
We tested the ability of human liver microsomes (HLMs) and recombinant human cytochrome P450 (CYP or P450) isoforms to catalyze the N-demethylation of nirvanol-free (S)-mephenytoin [(S)-MP] in vitro. In mixed HLMs, the kinetics of (S)-MP N-demethylation suggested two contributing activities.
R W Nims et al.
Journal of biochemical toxicology, 9(5), 269-278 (1994-10-01)
The abilities of structural congeners of phenobarbital to induce immunoreactive hepatic cytochrome P450 2B (CYP2B) protein and associated catalytic activity (benzyloxyresorufin O-dealkylation) in the male B6C3F1 mouse were examined. Interspecies differences in inducing ability were examined through comparison of the
R W Nims et al.
Journal of biochemical toxicology, 9(6), 279-288 (1994-12-01)
To explore the enantioselectivity of ligand interaction with the putative phenobarbital receptor, the pharmacodynamics of cytochrome P450 2B (CYP2B) induction by racemic 5-ethyl-5-phenylhydantoin and its two enantiomers were investigated in the male F344/NCr rat and in cultured adult male rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service