Key Documents
TMO382099-0250
Nalgene® Certified Low Particulate narrow-mouth bottle
natural high-density polyethylene, volume 250 mL, case of 72 ea
Synonym(s):
HDPE bottle, certified nalgene bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
natural high-density polyethylene
packaging
case of 72 ea
manufacturer/tradename
Nalgene 382099-0250
capacity
250 mL
volume
250 mL
General description
Effectively store high-purity chemicals with Thermo Scientific Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure, which are extremely low in trace metal content. Controlled environment manufacturing minimizes particulate contamination.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)