Skip to Content
Merck
All Photos(1)

Key Documents

T5398

Sigma-Aldrich

Transglutaminase from guinea pig liver

lyophilized powder, ≥1.5 units/mg protein

Synonym(s):

Protein-Glutamine-γ-Glutamyltransferase, Protein-glutamine:amine γ-glutamyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥1.5 units/mg protein

mol wt

76.6 kDa

composition

Protein, ≥80% modified Warburg-Christian

solubility

H2O: soluble 1.0 mg/mL, clear

application(s)

diagnostic assay manufacturing

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

This product from Sigma has been used to demonstrate that tissue transglutaminase (tTG) selectively deamidates gluten peptides, which results in strongly enhanced T cell-stimulatory activity. It has also been used to assess immune responses to A-gliadin peptides. Furthermore, it has been used to demonstrate that tTG selectively modifies gliadin peptides that are recognized by gut-derived T cells in celiac disease.
Transglutaminase has been used in a study to improve quantifiable assays to fully characterize the role of transglutaminase in diseases such as Huntington′s disease and Alzheimer′s disease.Transglutaminase has also been used in a study to develop a nonradioactive dot blot assay for transglutaminase activity.

Biochem/physiol Actions

Transglutaminase from guinea pig liver consists of a single polypeptide chain of 691 amino acid residues. It has six potential glycosylation sites (Asn-X-Ser or Asn-X-Thr), but it is not glycosylated. The molecular mass is approximately 76.6 kDa. It is calcium dependent and has several calcium binding sites. The enzyme is inhibited by iodoacetamide and N-ethylmaleimide in the presence of calcium. It catalyzes the incorporation of small molecular weight amines into γ-glutamine sites of proteins. In the absence of small molecular weight amines, it catalyzes the cross linking of proteins that results in the formation of γ-glutamyl-ε-lysine side chain peptides. Liver transglutaminase is a nonzymogenic enzyme.

Unit Definition

One unit will catalyze the formation of 1.0 μmole of hydroxamate per min from Nα-Z-Gln-Gly and hydroxylamine at pH 6.0 at 37 °C. (L-Glutamic acid γ-monohydroxamate is the standard.)

Physical form

Lyophilized powder containing Tris and dithioerythritol

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica C Kiefte-de Jong et al.
Gastroenterology, 144(4), 726-735 (2013-01-15)
Celiac disease in pregnant women has been associated with poor growth of the fetus, but little is known about how the level of celiac disease affects fetal growth or birth outcomes. We assessed the associations between levels of antibodies against
Syeda Warisul Fatima et al.
RSC advances, 11(55), 34613-34630 (2022-05-03)
Breast cancer is the most common malignancy among women. With the aim of decreasing the toxicity of conventional breast cancer treatments, an alternative that could provide appropriate and effective drug utilization was envisioned. Thus, we contemplated and compared the in
L Lorand et al.
Molecular and cellular biochemistry, 58(1-2), 9-35 (1984-01-01)
This paper is intended as a background to the topic of transglutaminases, while focusing on current ideas regarding the biological roles of these enzymes. Specifically, the following topics are discussed: geometry of forming gamma-glutamyl-epsilon-lysine cross-linked structures; energetic considerations; the gamma-glutamyl-epsilon-lysine
Mechanism of action of guinea pig liver transglutaminase. I. Purification and properties of the enzyme: identification of a functional cysteine essential for activity.
J E Folk et al.
The Journal of biological chemistry, 241(23), 5518-5525 (1966-12-10)
M P M Adriaanse et al.
Alimentary pharmacology & therapeutics, 37(4), 482-490 (2013-01-08)
Enterocyte damage is the hallmark of coeliac disease (CD) resulting in malabsorption. Little is known about the recovery of enterocyte damage and its clinical consequences. Serum intestinal fatty acid binding protein (I-FABP) is a sensitive marker to study enterocyte damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service