SRP0676
Sortase A, S. aureus, His-tag
Staphylococcus aureus, recombinant, N-terminal His-Tag
Synonym(s):
SrtA, sortase A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41202061
Recommended Products
recombinant
expressed in E. coli
tag
His tagged
form
liquid
mol wt
27.7 kDa
UniProt accession no.
storage temp.
−70°C
General description
Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
Application
This product is suitable for in vitro ligation of proteins and peptides to other. Sortase A has been shown to create circularized proteins [1], as well as couple polypeptides to a wide range of substituents including polymers [2], lipids [3], fluorophores [4], sugars [5], microspheres [3], peptide nucleic acids [6], among others.
Biochem/physiol Actions
Staphylococcal Sortase A is a bacterial transpeptidase that covalently links proteins to the bacterial cell wall by cleaving between threonine and glycine at an LPXTG recognition motif and catalyzes the formation of an amide bond between the carboxyl-group of threonine and the amino-group of the cell-wall peptidoglycan [7]. This chemistry can be exploited to site-specifically link proteins/peptides with the C-terminal LPETGX motif to other proteins or molecules possessing a glycine or aminomethylene motif [8].
Physical form
Aqueous buffered solution containing: 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 8 mM imidazole, 0.04% Tween-20, and 20% glycerol
Analysis Note
Analysis of this product can be performed in a reaction buffer (50 μl) containing 50 mM HEPES (pH=7.4), 150 mM NaCl, 5 mM CaCl2, 5 mM (Gly)3, 25 mM Abz/Dnp substrate, and Sortase A for 30 min at 30°C. Fluorescence intensity is measured at Ex320nm/Em420nm.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ranganath Parthasarathy et al.
Bioconjugate chemistry, 18(2), 469-476 (2007-02-17)
The Sortase family of transpeptidase enzymes catalyzes sequence-specific ligation of proteins to the cell wall of Gram-positive bacteria. Here, we describe the application of recombinant Staphylococcus aureus Sortase A to attach a tagged model protein substrate (green fluorescent protein) to
S K Mazmanian et al.
Science (New York, N.Y.), 285(5428), 760-763 (1999-07-31)
Surface proteins of Gram-positive bacteria are linked to the bacterial cell wall by a mechanism that involves cleavage of a conserved Leu-Pro-X-Thr-Gly (LPXTG) motif and that occurs during assembly of the peptidoglycan cell wall. A Staphylococcus aureus mutant defective in
Hongyuan Mao et al.
Journal of the American Chemical Society, 126(9), 2670-2671 (2004-03-05)
Sortase (SrtA), a transpeptidase from Staphylococcus aureus, catalyzes a cell-wall sorting reaction at an LPXTG motif by cleaving between threonine and glycine and subsequently joining the carboxyl group of threonine to an amino group of pentaglycine on the cell wall
John M Antos et al.
Journal of the American Chemical Society, 130(48), 16338-16343 (2008-11-08)
A general chemoenzymatic method for the site-specific attachment of lipids to protein substrates is described. Sortase A is used to append short lipid-modified oligoglycine peptides to the C terminus of protein substrates bearing a five amino acid sortase A recognition
Stephan Pritz et al.
The Journal of organic chemistry, 72(10), 3909-3912 (2007-04-17)
Sortase A is a transpeptidase that cleaves at a pentapeptide-motif and subsequently transfers the acyl component to a nucleophile containing N-terminal oligoglycines. We investigate the reaction conditions of the sortase-mediated ligation and demonstrate a useful application by the synthesis of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service