Skip to Content
Merck
All Photos(3)

Key Documents

T2151

Sigma-Aldrich

Glyceryl triheptadecanoate

≥99%

Synonym(s):

TG(17:0/17:0/17:0), 1,2,3-Triheptadecanoylglycerol, Triheptadecanoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)15COOCH2]2CHOCO(CH2)15CH3
CAS Number:
Molecular Weight:
849.40
Beilstein:
1811256
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCC

InChI

1S/C54H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-52(55)58-49-51(60-54(57)48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)50-59-53(56)47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h51H,4-50H2,1-3H3

InChI key

FBFWDBGUSMGXPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glyceryl triheptadecanoate has been used:
  • as an external standard in gas chromatography (GC) analysis for soybean seeds lipid extraction
  • as an internal standard solution to correct the transesterification efficiency during lipid analysis by gas chromatography-flame ionization detector (GC-FID)
  • as an internal standard in chloroform to dissolve total lipids obtained from fresh leaves of N. benthamiana for thin layer chromatography (TLC) analysis and to analyze their fatty acid content using GC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun Uk Kim et al.
FEBS open bio, 4, 25-32 (2013-12-24)
The LEAFY COTYLEDON2 (LEC2) gene plays critically important regulatory roles during both early and late embryonic development. Here, we report the identification of the LEC2 gene from the castor bean plant (Ricinus communis), and characterize the effects of its overexpression
Antoine Delbrut et al.
Molecules (Basel, Switzerland), 23(4) (2018-04-12)
By their autotrophic nature and their molecular richness, microalgae are serious assets in the context of current environmental and societal challenges. Some species produce both omega-3 long chain polyunsaturated fatty acids (PUFAs) and xanthophylls, two molecular families widely studied for
Kiran-Kumar Shivaiah et al.
Plant physiology, 182(2), 756-775 (2019-12-04)
Arabidopsis (Arabidopsis thaliana), like most dicotyledonous plants, expresses a multicomponent, heteromeric acetyl-CoA carboxylase (htACCase), which catalyzes the generation of the malonyl-CoA precursor of de novo fatty acid biosynthesis. This enzyme consists of four catalytic subunits: biotin carboxylase (BC), carboxyltransferase (CT)-α
Normand Podechard et al.
Scientific reports, 8(1), 7019-7019 (2018-05-08)
The extraction of RNA and lipids from a large number of biological samples is time-consuming and costly with steps required for both transcriptomic and lipidomic approaches. Most protocols rely on independent extraction of nucleic acids and lipids from a single
Shannon E Currie et al.
Conservation physiology, 7(1), coz005-coz005 (2019-02-26)
In order to effectively conserve species, we must understand the structure and function of integral mechanisms at all levels of organismal organisation, from intracellular biochemistry to whole animal ecophysiology. The accuracy of biochemical analyses depend on the quality and integrity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service