Skip to Content
Merck
All Photos(1)

Key Documents

T9126

Sigma-Aldrich

Glyceryl trioctanoate

≥95% (GC)

Synonym(s):

1,2,3-Tricapryloylglycerol, 1,2,3-Trioctanoylglycerol, Glycerol tricaprylate, Glycerol trioctanoate, Glyceryl tricaprylate, Tricaprylin, Trioctanoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)6COOCH2]2CHOCO(CH2)6CH3
CAS Number:
Molecular Weight:
470.68
Beilstein:
1717202
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

vegetable oil ( (coconut and palm kernel oils))

Quality Level

Assay

≥95% (GC)

form

liquid

bp

233 °C/1 mmHg (lit.)

density

0.956 g/mL at 20 °C (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCC(=O)OCC(COC(=O)CCCCCCC)OC(=O)CCCCCCC

InChI

1S/C27H50O6/c1-4-7-10-13-16-19-25(28)31-22-24(33-27(30)21-18-15-12-9-6-3)23-32-26(29)20-17-14-11-8-5-2/h24H,4-23H2,1-3H3

InChI key

VLPFTAMPNXLGLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glyceryl trioctanoate has been used:
  • as a solvent for quercetin dihydrate and used to study its healing effect on full thickness epidermal burn wound in Wistar
  • in the preparation of homogenous solution of silver complexes with different ligands to study its healing effect in a Wistar rat model of full-thickness burn
  • in the preparation of oil-water emulsions

Biochem/physiol Actions

Glyceryl trioctanoate might serve as a skin softening agent. It possesses caprylic acid as the aliphatic chain.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of emulsifier structure on lipid bioaccessibility in oil-water nanoemulsions
Speranza A, et al.
Journal of Agricultural and Food Chemistry, 61(26), 6505-6515 (2013)
The
Skin Care and Cosmetic Ingredients Dictionary, 180-180 (2014)
Healing effects of quercetin on full thickness epidermal thermal injury in Wistar rats.
Gouma E, et al.
International journal of phytomedicine, 277-281 (2016)
M K McKenna et al.
Cancer biology & therapy, 16(7), 1088-1098 (2015-05-29)
Withaferin A (WA), a withanolide from the plant, Ashwagandha (Withania somnifera) used in Ayurvedic medicine, has been found to be valuable in the treatment of several medical ailments. WA has been found to have anticancer activity against various solid tumors
Tehila Mishraki et al.
Colloids and surfaces. B, Biointerfaces, 75(1), 47-56 (2009-09-15)
A model protein (lysozyme) was incorporated into monoolein-based reverse hexagonal (H(II)) mesophase and its structure effects were characterized by small angle X-ray scattering, ATR-FTIR spectroscopy, and rheological measurements. Modifications in molecular organization of the H(II) mesophases as well as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service