Skip to Content
Merck
All Photos(1)

Key Documents

T1780

Sigma-Aldrich

Tyr-Tyr-Tyr-Tyr-Tyr-Tyr

≥98% (HPLC), suitable for mass spectrometry (MS)

Synonym(s):

Hexa-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H56N6O13
CAS Number:
Molecular Weight:
997.05
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Tyr-Tyr-Tyr-Tyr-Tyr-Tyr, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

composition

Peptide content, >60%

technique(s)

mass spectrometry (MS): suitable

color

white

storage temp.

−20°C

SMILES string

NC(Cc1ccc(O)cc1)C(=O)NC(Cc2ccc(O)cc2)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(Cc4ccc(O)cc4)C(=O)NC(Cc5ccc(O)cc5)C(=O)NC(Cc6ccc(O)cc6)C(O)=O

InChI

1S/C54H56N6O13/c55-43(25-31-1-13-37(61)14-2-31)49(67)56-44(26-32-3-15-38(62)16-4-32)50(68)57-45(27-33-5-17-39(63)18-6-33)51(69)58-46(28-34-7-19-40(64)20-8-34)52(70)59-47(29-35-9-21-41(65)22-10-35)53(71)60-48(54(72)73)30-36-11-23-42(66)24-12-36/h1-24,43-48,61-66H,25-30,55H2,(H,56,67)(H,57,68)(H,58,69)(H,59,70)(H,60,71)(H,72,73)

InChI key

JUUZKZKBBSFAAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J Buijs et al.
Journal of the American Society for Mass Spectrometry, 12(4), 410-419 (2001-04-27)
The isotopic exchange of amide hydrogens in proteins in solution strongly depends on the surrounding protein structure, thereby allowing structural studies of proteins by mass spectrometry. However, during electrospray ionization (ESI), gas phase processes may scramble or deplete the isotopic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service