Skip to Content
Merck
Tyr-Tyr-Tyr-Tyr-Tyr-Tyr ≥98% (HPLC)

Tyr-Tyr-Tyr-Tyr-Tyr-Tyr

Tyr-Tyr-Tyr-Tyr-Tyr-Tyr ≥98% (HPLC)
Synonym(s):
Hexa-L-tyrosine
Empirical Formula (Hill Notation):
C54H56N6O13
CAS Number:
Molecular Weight:
997.05

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing