Skip to Content
Merck
All Photos(1)

Key Documents

SRP3151

Sigma-Aldrich

SCF human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Steel Factor, Stem Cell Factor, c-Kit Ligand, Mast Cell Growth Factor (MGF)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤2.0 ng/mL ED50

mol wt

18.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KITLG(4254)

General description

SCF (stem cell factor) is a hematopoietic cytokine that exists in both soluble and transmembrane forms, which arise due to alternative splicing. Both these forms are functionally active. This gene is localized to human chromosome 12q22-12q24. Endothelial cells and fibroblasts constitutively produce both transmembrane and soluble forms of SCF. It is also produced by keratinocytes in normal skin, and epithelial cells in the gut, and thymus. Recombinant human SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.

Biochem/physiol Actions

SCF (stem cell factor) exerts its activity by signaling through the c-Kit receptor. They participate in hematopoiesis, and development of mast cells and melanocytes. Mutations in either of these proteins result in various phenotypic disorders, such as varying degrees of macrocytic anemia, suppressed numbers of tissue mast cells, reduced fertility, and decreased pigmentation. Linkage analysis along with whole-exome sequencing, c.286_303delinsT (p.Ser96Ter), shows a heterozygous truncating mutation in this gene linked with congenital and stable non-syndromic unilateral and asymmetric hearing loss. rs4590952 polymorphism in this gene is linked with disruption of p53 function through increase in p53-dependent SCF expression, which results in increased susceptibility to testicular cancer.

Sequence

MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA

Physical form

Lyophilized from 10 mM Acetic Acid.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A functional p53 responsive polymorphism in KITLG, rs4590952, does not affect the risk of breast cancer.
Chen W, et al.
Scientific Reports, 4:6371 (2014)
Stem cell factor and hematopoiesis.
Broudy VC
Blood, 90(4), 1345-1364 (1997)
Allelic Mutations of KITLG, Encoding KIT Ligand, Cause Asymmetric and Unilateral Hearing Loss and Waardenburg Syndrome Type 2.
Zazo Seco C, et al.
American Journal of Human Genetics, 97(5), 647-660 (2015)
T R Ulich et al.
Blood, 78(3), 645-650 (1991-08-01)
Recombinant rat stem cell factor (rrSCF) administered to rats as a single intravenous injection causes a dose-dependent neutrophilia and lymphocytosis as well as the appearance of immature myeloid cells and occasional blast cells in the circulation. Neutrophilia begins at 2
Sibylle A Brenner et al.
Immunology, 143(2), 174-183 (2014-04-05)
Mast cells are now considered sentinels in immunity. Given their location underneath the gastrointestinal barrier, mast cells are entrusted with the task of tolerating commensal microorganisms and eliminating potential pathogens in the gut microbiota. The aim of our study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service