Skip to Content
Merck
All Photos(1)

Key Documents

S7901

Sigma-Aldrich

Stem Cell Factor human

>97% (SDS-PAGE), recombinant, expressed in E. coli, powder, cell culture | mammalian: suitable

Synonym(s):

SCF, c-Kit ligand

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Stem Cell Factor human, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

powder

potency

1-5 ng/mL ED50

quality

endotoxin tested

mol wt

protein 18.5 kDa

packaging

pkg of 10 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μgtested (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... KITLG(4254)

Looking for similar products? Visit Product Comparison Guide

Application

Stem Cell Factor human has been used in
  • In vitro differentiation of equine embryonic stem cells
  • Chemotaxis detection system
  • Ex vivo expansion of umbilical cord blood CD34+ cells
  • Cytokine therapy

Biochem/physiol Actions

Epidermal stem cell factor (SCF) is considered as an essential mast cell growth factor and a multifunctional cytokine. SCF mediates tumor development and progression. Upregulation of the SCF gene is observed in hepatocellular carcinoma.
Stem Cell Factor (SCF), also known as c-Kit ligand (KL), steel factor (SLF) and mast cell growth factor (MGF), is a 30 kDa glycoprotein with broad activities on various tissues, including hematopoietic cells, pigment cells, and primordial germ cells.
Stem Cell Factor (SCF), also known as c-Kit ligand (KL), steel factor (SLF) and mast cell growth factor (MGF), is a 30 kDa glycoprotein with broad activities on various tissues, including hematopoietic cells, pigment cells, and primordial germ cells. SCF is secreted by endothelial cells, fibroblasts, and bone marrow stromal cells as a membrane-bound form which may be cleaved to release the soluble form. Both forms are active in promoting colony formation from murine bone marrow cells, but membrane-bound SCF is more effective in promoting hematopoieses in vivo, suggesting a role in cellular interactions between hematopoietic and stromal cells. The soluble is thought to exist in solution as a noncovalently linked dimer. SCF is structurally related to M-CSF (CSF-1) and Flt-3/Flk-2 Ligand (FL) with all three sharing a similar size, existence of transmembrane and soluble forms, four conserved cysteines, and alternative splicing exon locations, but they share little sequence homology. SCF alone is a modest colony stimulating factor. However, in the presence of other cytokines such as EPO, TPO, GM-CSF, G-CSF, M-CSF, IL-3, and IL-7, SCF is a potent costimulant that works synergistically to increase the size of myeloid, erythroid or lymphoid lineage colonies without influencing the lineage differentiation of the progenitors.

Physical form

Lyophilized from a 0.2 μm filtered solution in PBS containing 50 μg BSA per 1 μg as a carrier protein.

Analysis Note

The proliferative activity is tested in culture using TF1 human erythroleukemic cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto A Rovasio et al.
European journal of cell biology, 91(5), 375-390 (2012-03-03)
The extracellular environment through which neural crest cells (NCCs) translocate and differentiate plays a crucial role in the determination of cell migration and homing. In the trunk, NCC-derived melanocyte precursor cells (MPCs) take the dorsolateral pathway and colonize the skin
Isolation and ex vivo expansion of human umbilical cord blood-derived CD34+ stem cells and their cotransplantation with or without mesenchymal stem cells.
Delalat, B, et al.
Hematology, 14.3, 375-390 (2009)
Derivation, maintenance, and induction of the differentiation in vitro of equine embryonic stem cells
Saito S, et al.
Methods in Molecular Biology, 329, 59-79 (2006)
Combining pharmacological mobilization with intramyocardial delivery of bone marrow cells over-expressing VEGF is more effective for cardiac repair
Wang Y, et al.
Journal of Molecular and Cellular Cardiology, 40, 736-745 (2006)
Th17 cell-mediated immune responses promote mast cell proliferation by triggering stem cell factor in keratinocytes.
Cho KA, et al.
Biochemical and Biophysical Research Communications, 487(4), 856-861 (2017)

Articles

Read article on hematopoietic cytokines and hematopoiesis

Protocols

Stem cell reprogramming protocols to generate human induced pluripotent stem cells (iPSCs) including viral and non-viral RNA based methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service