SRP0335
Jak1 active human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)
Synonym(s):
JAK1A, JTK3, Janus kinase 1, jAK1B
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
34.1 kDa
packaging
pkg of 10 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... JAK1(3716)
General description
Human Jak1 (Janus kinase 1), also known as JTK3, (GenBank Accession No. NM_002227), amino acids 866-1155 (end) with N-terminal GST-Tag, MW= 59 kDa, expressed in Sf9 cells via a Baculovirus expression system.
JAK (Janus kinases) are activated when cytokines bind with their receptors, upon which they phosphorylate the receptors and produce docking sites for signaling molecules. One of the main signaling molecules includes signal transducer and activator of transcription (Stat) family. JAK1 is needed by multiple cytokines, including the family using the shared receptor subunit called common γ chain (γc), such as interleukin (IL)-2, IL-4, IL-7, IL-9, IL-15 and IL-21. JAK1 KO (knockout) mice have early postnatal lethal or perinatal lethal phenotype due to neurological defects. In vitro studies show that JAK1S703I mutation functions as an activator of JAK-STAT pathways even in the absence of cytokines, and drives cellular proliferation. JAK1S703I mutant patient-derived xenograft (PDX) HCC (hepatocellular carcinoma) cells also show sensitivity to JAK1/2 inhibitor, ruxolitinib.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Physical form
Formulated in 40 mM Tris-HCl pH 8.0, 110 mM NaCl, 2.2 mM KCl, 3 mM DTT,16 mM glutathione and 20% glycerol.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S J Rodig et al.
Cell, 93(3), 373-383 (1998-05-20)
Herein we report the generation of mice lacking the ubiquitously expressed Janus kinase, Jak1. Jak1-/- mice are runted at birth, fail to nurse, and die perinatally. Although Jak1-/- cells are responsive to many cytokines, they fail to manifest biologic responses
C Haan et al.
The Journal of biological chemistry, 276(40), 37451-37458 (2001-07-27)
Janus kinase 1 (Jak1) is a cytoplasmic tyrosine kinase that noncovalently associates with a variety of cytokine receptors. Here we show that the in vitro translated N-terminal domains of Jak1 are sufficient for binding to a biotinylated peptide comprising the
Activating JAK1 mutation may predict the sensitivity of JAK-STAT inhibition in hepatocellular carcinoma.
Yang S et al
Oncotarget, 7(5), 5461-5469 (2016)
T Miyazaki et al.
Science (New York, N.Y.), 266(5187), 1045-1047 (1994-11-11)
The interleukin-2 receptor (IL-2R) consists of three subunits: the IL-2R alpha, IL-2R beta, and IL-2R gamma chains, the last of which is also used in the receptors for IL-4, IL-7, and IL-9. Stimulation with IL-2 induces the tyrosine phosphorylation and
The Janus kinases (Jaks).
Yamaoka K et al
Genome Biology, 5(12), 253-253 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service