Skip to Content
Merck
All Photos(1)

Key Documents

SML2910

Sigma-Aldrich

Bindarit

≥98% (HPLC)

Synonym(s):

2-((1-Benzyl-1H-indazol-3-yl)methoxy)-2-methylpropanoic acid, 2-Methyl-2-[[1-(phenylmethyl)-1H-indazol-3-yl]methoxy]propanoic acid, AF 2838, AF2838

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O3
CAS Number:
Molecular Weight:
324.37
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C19H20N2O3/c1-19(2,18(22)23)24-13-16-15-10-6-7-11-17(15)21(20-16)12-14-8-4-3-5-9-14/h3-11H,12-13H2,1-2H3,(H,22,23)

InChI key

MTHORRSSURHQPZ-UHFFFAOYSA-N

Biochem/physiol Actions

Bindarit is an indazolic derivative with anti-inflammatory efficacy in vitro (150-300 μM) and in animal disease models in vivo (100 mg/kg ip, 50 mg/kg po, or 0.5% in chow; rats & mice). Bindarit selectively inhibits LPS-induced CCL2/MCP-1, CCL7/MCP-3, CCL8/MCP-2 chemokines over CCL3/MIP-1α and IL-8/CXCL8 in human monocytes, while blocking CCL2, but not CCL7 or CCL8, induction in HUVECs, and preventing LPS-induced CCL2 & IL-12β/p40 by inhibiting NFκB activation in murine macrophages (BMDM & Raw 264.7). Bindarit is also reported to exhibit non-competitive inhibiory potency against human monocarboxylate transporter MCT4 (Ki = 30.2 μM) over MCT1 (<40% inhibition at 500 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimiliano Mirolo et al.
European cytokine network, 19(3), 119-122 (2008-09-09)
Bindarit is an indazolic derivative that is devoid of any immunosuppressive effects and has no effect on arachidonic acid metabolism. However, it has been proved to have anti-inflammatory activity in a number of experimental diseases, including pancreatitis, arthritis, and lupus
Xiaoming Cai et al.
Nature communications, 9(1), 4416-4416 (2018-10-26)
Increasing concerns over the possible risks of nanotechnology necessitates breakthroughs in structure-activity relationship (SAR) analyses of engineered nanomaterials (ENMs) at nano-bio interfaces. However, current nano-SARs are often based on univariate assessments and fail to provide tiered views on ENM-induced bio-effects. Here we
A Guglielmotti et al.
Clinical and experimental rheumatology, 16(2), 149-154 (1998-04-16)
The present study was designed to investigate the effects of bindarit on animal survival and renal damage in murine lupus autoimmune disease. Female NZB/W mice were used. Bindarit was administered, as a 0.5% medicated diet, starting either before the onset
J L Steiner et al.
Cytokine, 66(1), 60-68 (2014-02-20)
Breast cancer, the most deadly cancer in women, is characterized by elevated levels of inflammation within and surrounding the tumor, which can lead to accelerated growth, invasion and metastasis. Macrophages are central to the inflammatory milieu and are recruited to
C Zoja et al.
Kidney international, 53(3), 726-734 (1998-03-21)
As an alternative to classical immunosuppressants in experimental lupus nephritis, we looked at bindarit, 2-methyl-2-[[1-phenylmethyl)-1H-indazol-3-y1]methoxy]propanoic acid, a novel molecule devoid of immunosuppressive effects, which selectively reduces chronic inflammation in rat adjuvant arthritis. Two groups of NZB/W mice (N = 55

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service