Skip to Content
Merck
All Photos(1)

Key Documents

SML2261

Sigma-Aldrich

Conivaptan hydrochloride

≥98% (HPLC)

Synonym(s):

N-[4-[(4,5-dihydro-2­methylimidazo[4,5-d][1]benzazepin-6(1H)-yl)carbonyl]phenyl]- [1,1’-biphenyl]-2-carboxamide monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H26N4O2 · HCl
CAS Number:
Molecular Weight:
535.04
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(C(N2CCC(N=C(C)N3)=C3C4=C2C=CC=C4)=O)C=C1)C5=CC=CC=C5C6=CC=CC=C6.Cl

InChI

1S/C32H26N4O2.ClH/c1-21-33-28-19-20-36(29-14-8-7-13-27(29)30(28)34-21)32(38)23-15-17-24(18-16-23)35-31(37)26-12-6-5-11-25(26)22-9-3-2-4-10-22;/h2-18H,19-20H2,1H3,(H,33,34)(H,35,37);1H

InChI key

BTYHAFSDANBVMJ-UHFFFAOYSA-N

Biochem/physiol Actions

Conivaptan hydrochloride is a novel dual Arginine vasopressin receptor (AVP-R) antagonist for AVP-R types 1a (V1a) and V2-R. Conivaptan potently inhibits AVP-induced intracellular signaling through human V2 and V1a receptors with no agonistic activity.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Tahara et al.
The Journal of pharmacology and experimental therapeutics, 282(1), 301-308 (1997-07-01)
The biochemical and pharmacological profile of YM087, 4'-[(2-methyl-1,4,5,6-tetrahydroimidazo[4,5-d][1]benzazepin- 6-yl)-carbonyl]-2-phenylbenzanilide monohydrochloride, a newly synthesized nonpeptide vasopressin (AVP) antagonist, was investigated in several in vitro and in vivo studies. YM087 showed high affinity for V1A receptors from rat liver and V2 receptors
Yukinori Arai et al.
Journal of pharmacological sciences, 109(1), 53-59 (2009-01-20)
Hyponatremia is the most common electrolyte disorder in hospitalized patients and is associated with the risk of intractable seizures and death. The effectiveness of conventional therapies for hyponatremia is inconsistent, and the rapid correction of plasma sodium levels is thought
Poornima Vinod et al.
Cardiology research, 8(3), 87-95 (2017-07-21)
Heart and kidney failure continued to be of increasing prevalence in today's society, and their comorbidity has synergistic effect on the morbidity and mortality of patients. Cardiorenal syndrome (CRS) is a complex disease with multifactorial pathophysiology. Better understanding of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service