SML2805
Narciclasine
≥98% (HPLC)
Synonym(s):
Narciclasine, (2S-(2-alpha,3-beta,4-beta,4a-beta))-3,4,4a,5-tetrahydro-2,3,4,7-tetrahydroxy-(1,3)Dioxolo(4,5-j)phenanthridin-6(2H)-one, 3,4,4a,5-Tetrahydro-2,3,4,7-tetrahydroxy-(1,3)dioxolo(4,5-j)phenanthridin-6(2H)-one, BRN 1087400, Lycoricidin-A, Lycoricidinol, NSC 266535
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
optical activity
[α]/D 167 to 187° in DMSO
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
−20°C
SMILES string
O[C@H]1C=C2[C@@H](NC(=O)c3c(O)c4OCOc4cc23)[C@H](O)[C@@H]1O
InChI
1S/C14H13NO7/c16-6-1-5-4-2-7-13(22-3-21-7)11(18)8(4)14(20)15-9(5)12(19)10(6)17/h1-2,6,9-10,12,16-19H,3H2,(H,15,20)/t6-,9+,10+,12-/m0/s1
InChI key
LZAZURSABQIKGB-AEKGRLRDSA-N
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | PHL82604 | SML2620 | SML2326 |
---|---|---|---|
≥98% (HPLC) | ≥98.0% (HPLC) | ≥98% (HPLC) | ≥98% (HPLC) |
100 | - | - | - |
−20°C | - | 2-8°C | 2-8°C |
DMSO: 2 mg/mL, clear | - | DMSO: 2 mg/mL, clear | DMSO: 2 mg/mL, clear |
white to beige | - | faint yellow to dark orange | white to beige |
Biochem/physiol Actions
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service