Skip to Content
Merck
All Photos(1)

Key Documents

SML2678

Sigma-Aldrich

A-770041

≥98% (HPLC)

Synonym(s):

A 770041, A770041, N-[4-[1-[trans-4-(4-Acetyl-1-piperazinyl)cyclohexyl]-4-amino-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-2-methoxyphenyl]-1-methyl-1H-indole-2-carboxamide, trans-KIN 001-111

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H39N9O3
CAS Number:
Molecular Weight:
621.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1C(C3=CC=C(NC(C4=CC(C=CC=C5)=C5N4C)=O)C(OC)=C3)=NN2[C@@H]6CC[C@@H](N7CCN(C(C)=O)CC7)CC6

InChI

1S/C34H39N9O3/c1-21(44)41-14-16-42(17-15-41)24-9-11-25(12-10-24)43-33-30(32(35)36-20-37-33)31(39-43)23-8-13-26(29(19-23)46-3)38-34(45)28-18-22-6-4-5-7-27(22)40(28)2/h4-8,13,18-20,24-25H,9-12,14-17H2,1-3H3,(H,38,45)(H2,35,36,37)

InChI key

ZMNWFTYYYCSSTF-UHFFFAOYSA-N

Biochem/physiol Actions

A-770041 is an orally available (rat F = 34.1%; 10 mg/kg p.o.), ATP-competitive, potent and selctive Lck inhibtor (IC50 = 147 nM/Lck, 1.18 μM/Lyn, 1.22 μM/Hck, 9.05 μM/Src, 14.1 μM/Fgr, 44.1 μM/Fyn; >25 μM/IKK1, IKK2, IRAK4, p38, JAK3, ZAP70, ITK, PKC, MK2, PKA, TYK2, COT,Syk; >50 μM/Tie-2 & Kdr; 1 mM ATP). A-770041 inhibits CD3 mAb-induced IL-2 production in human whole blood in vitro (IC 50 = 80 nM) and in mice in vivo (IC50 = 78 nM plasma conc.), and significantly lengthens graft survival time in a rat heterotopic heart transplant model (2.5 & 5 mg/kg bid p.o.).
Orally available, ATP-competitive, potent and selctive Lck inhibtor with immune suppression efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Jing Wu et al.
Frontiers in pharmacology, 9, 4-4 (2018-02-08)
Researchers have shown that the level of immunoglobulin D (IgD) is often elevated in patients with autoimmune diseases. The possible roles of IgD on the function of human T cell activation are still unclear. Paeoniflorin-6'-O-benzene sulfonate (code: CP-25), the chemistry
Andrew Burchat et al.
Bioorganic & medicinal chemistry letters, 16(1), 118-122 (2005-10-12)
We describe the identification, SAR, and pharmacology of the src-family selective lck inhibitor A-770041 that prolongs the survival of major histocompatibility mismatched allografts in models of solid organ transplant rejection for greater than 65 days.
J P Zepecki et al.
Oncogene, 38(10), 1734-1750 (2018-10-26)
Migration of human glioma cells (hGCs) within the brain parenchyma makes glioblastoma one of the most aggressive and lethal tumors. Studies of the cellular and molecular mechanisms underlying hGC migration are hindered by the limitations of existing glioma models. Here
X Li et al.
Allergy, 73(2), 361-368 (2017-09-01)
In mast cells, induction of HSP70 expression during antigen stimulation has not been reported. Mouse bone marrow-derived mast cells (BMMC) were stimulated with IgE/Ag or HSP70. Induction of HSP70 expression and signaling protein phosphorylation were evaluated by immunoblotting. HSP70 expression
Zhenfeng Duan et al.
BMC cancer, 14, 681-681 (2014-09-23)
Reversing multidrug resistance (MDR) has been an important goal for clinical and investigational oncologists. In the last few decades, significant effort has been made to search for inhibitors to reverse MDR by targeting ATP-binding cassette (ABC) transporters (Pgp, MRP) directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service