Skip to Content
Merck
All Photos(4)

Key Documents

A2385

Sigma-Aldrich

5-Azacytidine

≥98% (HPLC), powder, DNA methyltransferase inhibitor

Synonym(s):

5-Azacitidine, 4-Amino-1-(β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, Ladakamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O5
CAS Number:
Molecular Weight:
244.20
Beilstein:
620461
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

5-Azacytidine, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

mp

226-232 °C (dec.) (lit.)

antibiotic activity spectrum

viruses

Mode of action

DNA synthesis | interferes

originator

Celgene

storage temp.

−20°C

SMILES string

NC1=NC(=O)N(C=N1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

InChI

1S/C8H12N4O5/c9-7-10-2-12(8(16)11-7)6-5(15)4(14)3(1-13)17-6/h2-6,13-15H,1H2,(H2,9,11,16)/t3-,4-,5-,6-/m1/s1

InChI key

NMUSYJAQQFHJEW-KVTDHHQDSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

5-Azacytidine is a chemical analog of cytidine. It functions as a hypomethylating agent. 5-Azacytidine is used to treat myelodysplastic syndrome (MDS).
Chemical structure: nucleoside

Application

5-Azacytidine has been used:
  • for cell induction
  • to study its effects on bovine fetal mesenchymal stem cells (bfMSC)
  • to study its effects on human bone marrow stromal cells (hBMSCs)
  • to study its effects on and DNA methyltransferase 1 (DNMT1) and Ras protein activator like 1 (RASAL1) expression
  • to interfere with DNA methylation and histone acetylation
  • to determine its effects on the conversion of control fibroblasts
  • for optical coherence tomography (OCT) and fluorescein angiography (FA)
  • for the reactivation of Sal-like protein (SALL)3 expression

Biochem/physiol Actions

5-Azacytidine (Aza-CR) acts as a potential chemotherapeutic regimen for acute myelogenous leukemia. This drug has an ability to selectively increase γ-globin synthesis. Therefore, 5-azacytidine is used in treating severe β-thalassemia. Aza-CR acts as a potential bacteriostatic, antitumor and mutagenic agent. In addition, it also exhibits various biological activity such as, immunosuppressive, antimitotic, radioprotective and virostatic effects.
A potent growth inhibitor and cytotoxic agent; inhibits DNA methyltransferase, an important regulatory mechanism of gene expression, gene activation and silencing.
Causes DNA demethylation or hemi-demethylation, creating openings that allow transcription factors to bind to DNA and reactivate tumor suppressor genes.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Celgene. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional striated muscle cells from non-myoblast precursors following 5-azacytidine treatment
Constantinides PG, et al.
Nature, 267(5609), 364-364 (1977)
Myogenic differentiation potential of mesenchymal stem cells derived from fetal bovine bone marrow
Okamura LH, et al.
Animal Biotechnology, 29(1), 1-11 (2018)
Dexamethasone regulates EphA5, a potential inhibitory factor with osteogenic capability of human bone marrow stromal cells
Yamada T, et al.
Stem Cells International, 2016 (2016)
Enhanced conversion of induced neuronal cells (iN cells) from human fibroblasts: utility in uncovering cellular deficits in mental illness-associated chromosomal abnormalities
Passeri E, et al.
Neuroscience Research, 101, 57-61 (2015)
5-Azacytidine selectively increases gamma-globin synthesis in a patient with beta thalassemia
Ley TJ and DeSimone, et al.
The New England Journal of Medicine, 307(24), 1469-1475 (1982)

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service