Skip to Content
Merck
All Photos(1)

Key Documents

SML2331

Sigma-Aldrich

Perospirone hydrochloride

≥98% (HPLC)

Synonym(s):

N-[4-[4-(1,2-benzisothiazol-3-yl)-1-piperazinyl]butyl]-1,2-cis-cyclohexanedicarboximide hydrochloride, SM-9018 hydrochloride, rel-(3aR,7aS)-2-[4-[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]butyl]hexahydro-1H-isoindole-1,3(2H)-dione hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N4O2S · HCl
CAS Number:
Molecular Weight:
463.04
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C23H30N4O2S.ClH/c28-22-17-7-1-2-8-18(17)23(29)27(22)12-6-5-11-25-13-15-26(16-14-25)21-19-9-3-4-10-20(19)30-24-21;/h3-4,9-10,17-18H,1-2,5-8,11-16H2;1H/t17-,18+;

InChI key

HIZFAPMOZFYELI-GNXQHMNLSA-N

Biochem/physiol Actions

Perospirone (SM-9018) is a typical neuroleptic (antipsychotic) agent that displays high affinity for 5-HT2, D2 and 5-HT1A receptors (Ki = 0.61, 1.4 and 2.9-1.8 nM, respectively), moderate affinity for alpha 1 and D1 receptors (Ki = 17 and 41 nM, respectively), while exhibiting much reduced affinity for alpha 2 (Ki = 408 nM) and little affinity toward muscarinic, opiate, glutamate, phencyclidine, benzodiazepine or GABAA receptors (Ki >1 μM). SM-9018 antagonizes both D2-dependent (e.g. methamphetamine-induced hyperactivity, apomorphine-induced stereotypy) and 5-HT2-mediated (e.g. tryptamine-induced clonic seizure) activities in vivo, while exhibiting very low cataleptogenic and central depressant activities commonly observed with two other neuroleptics, haloperidol and chlorpromazine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Ohno et al.
Pharmacology, biochemistry, and behavior, 49(1), 19-23 (1994-09-01)
Induction of bradykinesia by SM-9018, a novel 5-HT2 and D2 antagonist, was compared with that of other neuroleptics using the pole test in mice. Neuroleptics including SM-9018, haloperidol, chlorpromazine, and thioridazine dose dependently induced bradykinesia in the pole-descending behavior of
A Hirose et al.
Japanese journal of pharmacology, 53(3), 321-329 (1990-07-01)
Pharmacological studies were undertaken to clarify the profile of cis-2-(4-(4-(1,2-benzisothiazol-3-yl)-1-piperazinyl) butyl) hexahydro-1H-isoindole-1,3-(2H)-dione hydrochloride (SM-9018), a new neuroleptic drug. SM-9018 had very high binding affinities for both 5-hydroxytryptamine2 (5-HT2) and dopamine2 (D2) receptors, unlike many other neuroleptics. SM-9018 also strongly inhibited
Y Ohno et al.
General pharmacology, 26(3), 489-494 (1995-05-01)
1. Changes in behavioral dopaminergic and serotonergic sensitivities were studied in rats after withdrawal of the chronic treatments with SM-9018 (0.1 mg/kg/day), a potential atypical neuroleptic, and with haloperidol (0.3 mg/kg/day) using continuous infusion pumps. 2. Administration of SM-9018 inhibited
Y Ohno et al.
Progress in neuro-psychopharmacology & biological psychiatry, 19(6), 1091-1101 (1995-10-01)
1. Receptor binding and behavioral studies were performed to compare the effects of subchronic treatments with SM-9018, a novel 5-HT2 and D2 antagonist, and with haloperidol (HAL) on dopamine and 5-HT receptors in rats. 2. SM-9018 treatment (10 mg/kg/day p.o.)
T Kato et al.
Japanese journal of pharmacology, 54(4), 478-481 (1990-12-01)
The present study employed various receptor-binding assays to clarify the biochemical characteristics of SM-9018. SM-9018 possessed very high affinity for 5-HT2, D2 and 5-HT1A receptors (Ki = 0.61, 1.4 and 2.9 nM, respectively), and it had moderate affinity for alpha

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service