Skip to Content
Merck
All Photos(3)

Key Documents

M9020

Sigma-Aldrich

Mecamylamine hydrochloride

≥98% (TLC), powder, nAChR antagonist

Synonym(s):

2-(Methylamino)isocamphane hydrochloride, Inversine, N,2,3,3-Tetramethylbicyclo[2.2.1]heptan-2-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21N · HCl
CAS Number:
Molecular Weight:
203.75
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

product name

Mecamylamine hydrochloride,

form

powder

Quality Level

solubility

ethanol: 122 mg/mL
H2O: 47 mg/mL
isopropanol: 476 mg/mL
glycerol: 95 mg/mL

originator

AstraZeneca

SMILES string

Cl.[H][C@@]12CC[C@@]([H])(C1)C(C)(NC)C2(C)C

InChI

1S/C11H21N.ClH/c1-10(2)8-5-6-9(7-8)11(10,3)12-4;/h8-9,12H,5-7H2,1-4H3;1H/t8-,9+,11?;/m1./s1

InChI key

PKVZBNCYEICAQP-CIISUUNXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mecamylamine hydrochloride has been used as an additive in extracellular saline during current-clamp recordings to reduce synaptic input. It has also been used as a non-selective nicotinic acetyl choline receptor blocker in aortic body neurons and in MLO-Y4 cells.

Biochem/physiol Actions

Noncompetitive nicotinic acetylcholine receptor antagonist; preferentially blocks nicotinic receptors at autonomic ganglia; crosses blood-brain barrier.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

Solutions are stable to autoclaving.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetylcholine affects osteocytic MLO-Y4 cells via acetylcholine receptors
Ma Y, et al.
Molecular and cellular endocrinology, 384(1-2), 155-164 (2014)
M I Damaj et al.
The Journal of pharmacology and experimental therapeutics, 291(3), 1284-1291 (1999-11-24)
Pharmacological mechanisms involved in nicotine-induced seizures were investigated in mice by testing the ability of several nicotinic agonists in producing seizures after peripheral administration. In addition, nicotinic antagonists such as hexamethonium, mecamylamine, dihydro-beta-erythroidine, and methyllycaconitine citrate (MLA) were used in
G Panagis et al.
Synapse (New York, N.Y.), 35(1), 15-25 (1999-12-01)
In the present study the neuronal expression of Fos, the protein product of c-fos, was used to study changes in neuronal activity in nerve terminal regions of the ascending dopaminergic system during nicotine withdrawal. Rats were infused for 14 days
Potential roles of ATP and local neurons in the monitoring of blood O2 content by rat aortic bodies
Piskuric NA, et al.
Experimental Physiology, 99(1), 248-261 (2014)
Olena Riabinina et al.
Genetics, 212(1), 53-63 (2019-03-14)
The Q-system is a binary expression system that works well across species. Here, we report the development and demonstrate the applications of a split-QF system that drives strong expression in Drosophila, is repressible by QS, and is inducible by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service