Skip to Content
Merck
All Photos(1)

Key Documents

SML2321

Sigma-Aldrich

Gestrinone

≥97% (HPLC)

Synonym(s):

(17α)-13-Ethyl-17-hydroxy-18,19-dinorpregna-4,9,11-trien-20-yn-3-one, Ethylnorgestrienone, R-2323, R2323, RU-2323

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O2
CAS Number:
Molecular Weight:
308.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

optical activity

[α]/D +81 to +88°, c = 0.4 in ethanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@@]1(C#C)CC[C@@]2([H])[C@]3([H])CCC4=CC(CCC4=C3C=C[C@@]21CC)=O

InChI

1S/C21H24O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,9,11,13,18-19,23H,3,5-8,10,12H2,1H3/t18-,19+,20+,21+/m1/s1

InChI key

BJJXHLWLUDYTGC-ANULTFPQSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gestrinone is a synthetic steroid, an analog of norgestrienone, which exhibits androgenic, anti-estrogenic and anti-progestogenic activities. Gestrinone is used for treatment of endometriosis.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison K Death et al.
The Journal of clinical endocrinology and metabolism, 89(5), 2498-2500 (2004-05-06)
Tetrahydrogestrinone (THG) was recently identified as a novel steroid used illicitly to improve athletic performance. Although its structure is closely related to gestrinone, a 19-nor progestin, and resembles that of trenbolone, THG was never marketed, so information on its hormonal
Yan Zhu et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 66(8), 569-577 (2012-10-30)
The study was to investigate the effect of gestrinone on the growth of human uterine leiomyoma cells and on the levels and activity of p38, Src and estrogen receptor alpha (ERα). Human uterine leiomyoma cells were cultured and treated with
T Tamaya et al.
Acta obstetricia et gynecologica Scandinavica, 65(5), 439-441 (1986-01-01)
To understand the mechanism of biological action of gestrinone (R2323), which has a therapeutic effect against endometriosis, the binding of gestrinone to numerous classes of intracellular steroid binding proteins was studied in the human uterine endometrium. Gestrinone bound to endometrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service