Skip to Content
Merck
All Photos(4)

Key Documents

T1875

Sigma-Aldrich

Testosterone propionate

solid

Synonym(s):

17β-Hydroxy-4-androsten-3-one 17-propionate, 17β-Propionyloxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one 17-propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
Beilstein:
3221760
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

solid

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

solubility

acetone: soluble 1 in 4 of acetone
chloroform: soluble 50 mg/ml, clear, colorless to faintly yellow
ethanol: soluble 1 in 6 of ethanol
ethyl oleate: soluble 1 in 20 of ethyl oleate
propylene glycol: soluble 1 in 30 of propylene glycol
soluble 1 in 20 of ethyl oleate
ethanol: 10 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 4.4 mg/mL
H2O: insoluble
oil: soluble

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])OC(=O)CC

InChI

1S/C22H32O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h13,16-19H,4-12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1

InChI key

PDMMFKSKQVNJMI-BLQWBTBKSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Testosterone propionate has been used for studying its effects on pregnant ewes,.

Biochem/physiol Actions

Androgens direct the development of the male phenotype during embryogenesis and at puberty. Testosterone is an androgen that is secreted by the testis. This hormone is converted to dihydrotestosterone in the target tissues where it regulates several biological functions. Testosterone propionate has been synthetically derived from a plant. This product has extended and faster-acting functions when compared to other testosterone esters.

Physical form

Photosensitive solid

Preparation Note

This product is soluble in chloroform (50 mg/ml). It is also soluble in various oils. The oil solutions can be sterilized by maintaining at 150 deg C for 1 hour. It is practically insoluble in water, soluble 1 in 6 of ethanol, 1 in 4 of acetone, 1 in 20 of ethyl oleate, and 1 in 30 of propylene glycol. The product is incompatible with alkalis and oxidizing agents and it should be protected from light.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M E Rose et al.
Infection and immunity, 26(2), 630-637 (1979-11-01)
Comparisons were made between infections with Eimeria spp. in normal animals and in animals with functional deficiencies in either T-lymphocytes (athymic nude rats) or B-lymphocytes (bursectomized chickens). Approximately three times more oocysts of E. nieschulzi were passed during a primary
Maria Cernea et al.
Endocrinology, 156(9), 3277-3291 (2015-06-11)
Prenatal testosterone (T)-treated ewes display a constellation of reproductive defects that closely mirror those seen in PCOS women, including altered hormonal feedback control of GnRH. Kisspeptin/neurokinin B/dynorphin (KNDy) neurons of the arcuate nucleus (ARC) play a key role in steroid
Tsuyoshi Shimmura et al.
Scientific reports, 9(1), 3978-3978 (2019-03-10)
Animals that communicate using sound are found throughout the animal kingdom. Interestingly, in contrast to human vocal learning, most animals can produce species-specific patterns of vocalization without learning them from their parents. This phenomenon is called innate vocalization. The underlying
Mohan Manikkam et al.
Biology of reproduction, 78(4), 648-660 (2007-12-21)
The goal of this study was to explore mechanisms that mediate hypersecretion of LH and progressive loss of cyclicity in female sheep exposed during fetal life to excess testosterone. Our working hypothesis was that prenatal testosterone excess, by its androgenic
Carlo Serra et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 68(1), 17-26 (2012-04-14)
Aging is associated with loss of muscle mass and strength, reduced satellite cell number, and lower regenerative potential. Testosterone increases muscle mass, strength, and satellite cell number in humans; however, the effects of testosterone on the regenerative potential of skeletal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service