Skip to Content
Merck
All Photos(1)

Key Documents

SML0720

Sigma-Aldrich

Methazolamide

≥98% (HPLC)

Synonym(s):

5-Acetylimino-4-methyl-·2-1,3,4-thiadiazoline-2-sulfonamide, N-(4-Methyl-2-sulfamoyl-Δ2-1,3,4-thiadiazolin-5-ylidene) acetamide, L 584601, N-[5-(Aminosulfonyl)-3-methyl-1,3,4-thiadiazol-2(3H)-ylidene]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N4O3S2
CAS Number:
Molecular Weight:
236.27
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

mp

208 °C (dec.) (lit.)

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1N=C(S\C1=N\C(C)=O)S(N)(=O)=O

InChI

1S/C5H8N4O3S2/c1-3(10)7-4-9(2)8-5(13-4)14(6,11)12/h1-2H3,(H2,6,11,12)/b7-4+

InChI key

FLOSMHQXBMRNHR-QPJJXVBHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cell-permeable and potent carbonic anhydrase (CA) inhibitor
Methazolamide has a plasma half-life of 14 hours in humans. Methazolamide has relatively good lipid solubility and low plasma protein binding ability. Thus, it diffuses into tissues and fluids much more readily than acetazolamide, another carbonic anhydrase inhibitor. Methazolamide has less side effects than acetazolamide.
Methazolamide is a cell-permeable and potent carbonic anhydrase (CA) inhibitor that is used in the treatment of glaucoma. Methazolamide is an insulin sensitizer that reduces hepatic glucose generation in animal models.

Other Notes

Light sensitive

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The pharmacology of methazolamide in relation to the treatment of glaucoma.
Maren TH, et al.
Investigative Ophthalmology & Visual Science, 16(8), 730-742 (1977)
Ming-Tao Yang et al.
Psychopharmacology, 232(20), 3763-3772 (2015-08-01)
Dysregulation of noradrenergic and dopaminergic systems is involved in the pathology of attention deficit hyperactivity disorder (ADHD). Carbonic anhydrase (CA) has been reported to affect monoamine transmission in the central nervous system. The aim of this study is to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service