Skip to Content
Merck
All Photos(1)

Key Documents

SML1414

Sigma-Aldrich

Liproxstatin-1

>98% (HPLC), powder, ferroptosis inhibitor

Synonym(s):

N-[(3-Chlorophenyl)methyl]-spiro[piperidine-4,2′(1′H)-quinoxalin]-3′-amine, N-[(3-chlorophenyl)methyl]spiro[4H-quinoxaline-3,4′-piperidine]-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21ClN4
CAS Number:
Molecular Weight:
340.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Liproxstatin-1, >98% (HPLC)

Quality Level

Assay

>98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC(CNC2=NC3=CC=CC=C3NC24CCNCC4)=CC=C1

InChI

1S/C19H21ClN4/c20-15-5-3-4-14(12-15)13-22-18-19(8-10-21-11-9-19)24-17-7-2-1-6-16(17)23-18/h1-7,12,21,24H,8-11,13H2,(H,22,23)

InChI key

YAFQFNOUYXZVPZ-UHFFFAOYSA-N

Application

Liproxstatin-1 has been used as a cell death inhibitor in the cell viability assay and for the determination of lipid peroxidation.

Biochem/physiol Actions

Liproxstatin-1 is a potent inhibitor of ferroptosis, a non-apoptotic form of cell death characterized by iron-dependent accumulation of lethal lipid reactive oxygen species (ROS). Liproxstatin-1 suppressed ferroptosis in human cells and in an ischaemia/reperfusion-induced tissue injury model in mice. Knockout of glutathione peroxidase 4 (Gpx4) Has been shown to cause cell death by ferroptosis. Liproxstatin-1 was able to suppress ferroptosis in Gpx4 knock-out mice.
Liproxstatin-1 is a potent inhibitor of ferroptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selenium utilization by GPX4 is required to prevent hydroperoxide-induced ferroptosis.
Ingold I, et al.
Cell, 172(3), 409-422 (2018)
Alejandra M Martinez et al.
FEBS open bio, 9(4), 582-593 (2019-04-16)
Ferroptosis is a form of regulated cell death that is driven by lethal accumulation of lipid peroxides upon inhibition of glutathione peroxidase 4 (GPx4). Deletion of the Gpx4 gene in mice revealed that neurons are sensitive to ferroptosis in vivo. However
Ji-Yoon Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(51), 32433-32442 (2020-12-09)
Ferroptosis is an iron-dependent regulated necrosis mediated by lipid peroxidation. Cancer cells survive under metabolic stress conditions by altering lipid metabolism, which may alter their sensitivity to ferroptosis. However, the association between lipid metabolism and ferroptosis is not completely understood.
Masahiro Yoshida et al.
Nature communications, 10(1), 3145-3145 (2019-07-19)
Ferroptosis is a necrotic form of regulated cell death (RCD) mediated by phospholipid peroxidation in association with free iron-mediated Fenton reactions. Disrupted iron homeostasis resulting in excessive oxidative stress has been implicated in the pathogenesis of chronic obstructive pulmonary disease
Yilong Zou et al.
Nature, 585(7826), 603-608 (2020-09-18)
Ferroptosis-an iron-dependent, non-apoptotic cell death process-is involved in various degenerative diseases and represents a targetable susceptibility in certain cancers1. The ferroptosis-susceptible cell state can either pre-exist in cells that arise from certain lineages or be acquired during cell-state transitions2-5. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service