Skip to Content
Merck
All Photos(1)

Key Documents

480065

Sigma-Aldrich

Necrostatin-1

≥95% (HPLC), crystalline solid, necroptosis inhibitor, Calbiochem

Synonym(s):

Necrostatin-1, Nec-1, Necrosome Inhibitor I, 5-(Indol-3-ylmethyl)-(2-thio-3-methyl)hydantoin, Necrosis Inhibitor II, Methyl-thiohydantoin-tryptophan, MTH-Trp, Receptor-Interacting Protein 1 Inhibitor I, RIPK 1 Inhibitor I, Nec1, RIP1 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3OS
CAS Number:
Molecular Weight:
259.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Necrostatin-1, Necrostatin-1, CAS 4311-88-0, is a cell-permeable, potent, and selective blocker of necroptosis (EC₅₀ = 494 nM in FADD-deficient Jurkat cells treated with TNF-α).

Quality Level

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C13H13N3OS/c1-16-12(17)11(15-13(16)18)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,11,14H,6H2,1H3,(H,15,18)

InChI key

TXUWMXQFNYDOEZ-UHFFFAOYSA-N

General description

A cell-permeable, potent, and selective blocker of necroptosis (EC50 = 494 nM in FADD-deficient Jurkat cells treated with TNF-α), a nonapoptotic necrotic cell death pathway mediated by death-domain receptors (DRs) that offers neuroprotection in a murine model of ischemic brain injury. Exhibits no effect on DR-induced apoptosis. Also acts as a selective and ATP-competitive inhibitor of RIP1 kinase with negligible effect of RIP2 kinase activity. Nec-1 target appears to be a critical common necroptotic step upstream of execution events and downstream of DRs. Inactive control, Nec-1i, is also available (Cat. No. 480066).
A cell-permeable, potent, and selective blocker of necroptosis (EC50 = 494 nM in FADD-deficient Jurkat cells treated with TNF-α), a nonapoptotic necrotic cell death pathway mediated by death-domain receptors (DRs) that offers neuroprotection in a murine model of ischemic brain injury. Exhibits no effect on DR-induced apoptosis. Also acts as a selective and ATP-competitive inhibitor of RIP1 kinase with negligible effect of RIP2 kinase activity. Nec-1 target appears to be a critical common necroptotic step upstream of execution events and downstream of DRs. Inactive control, Nec-1i, is also available (Cat. No. 480066). Also available as a 25 mM solution in DMSO (Cat. No. 505224).

Biochem/physiol Actions

Cell permeable: yes
EC50 = 494 nM in blocking necrosis in FADD-deficient Jurkat cells treated with TNF-α
Primary Target
Blocker of necroptosis
Product competes with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Degterev, A., et al. 2013. Nat. Chem. Biol.9, 192.
Degterev, A., et al. 2012. Cell Death Differ.20, 366.
Christofferson, D.E., et al. 2012. Cell Death Dis.3, e320.
Takahashi, N., et al. 2012. Cell Death Dis.3, e437.
Degterev, A., et al. 2008. Nat. Chem. Biol.4, 313.
Degterev, A., et al. 2005. Nat. Chem. Biol.1, 112.
Muller, A.J., et al. 2005. Nat. Med.11, 312.
Teng, X., et al. 2005. Bioorg. Med. Chem. Lett.15, 5039.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Fernández-Acosta et al.
Molecules (Basel, Switzerland), 27(13) (2022-07-10)
The use of nanomaterials rationally engineered to treat cancer is a burgeoning field that has reported great medical achievements. Iron-based polymeric nano-formulations with precisely tuned physicochemical properties are an expanding and versatile therapeutic strategy for tumor treatment. Recently, a peculiar
Bolin Hou et al.
Cell death discovery, 8(1), 319-319 (2022-07-14)
The underlying mechanism by which growth factor receptor-bound protein 2 (Grb2) regulates necroptosis remains unexplored. In the present study, we found that rasfonin, a fungal natural product and an activator of necroptosis, enhanced Grb2 binding to receptor-interacting serine/threonine kinase 1
Sasker Grootjans et al.
Nature protocols, 11(8), 1444-1454 (2016-07-16)
Several cell death assays have been developed based on a single biochemical parameter such as caspase activation or plasma membrane permeabilization. Our fluorescent apoptosis/necrosis (FAN) assay directly measures cell death and distinguishes between caspase-dependent apoptosis and caspase-independent necrosis of cells
Peng He et al.
Cell death discovery, 10(1), 255-255 (2024-05-25)
Caspase-8 (Casp8) serves as an initiator of apoptosis or a suppressor of necroptosis in context-dependent manner. Members of the p90 RSK family can phosphorylate caspase-8 at threonine-265 (T265), which can inactivate caspase-8 for bypassing caspase-8-mediated blockade of necroptosis and can
Kana Otsubo et al.
FEBS letters, 594(10), 1586-1595 (2020-01-31)
Autophagy is an intracellular process that regulates the degradation of cytosolic proteins and organelles. Dying cells often accumulate autophagosomes. However, the mechanisms by which necroptotic stimulation induces autophagosomes are not defined. Here, we demonstrate that the activation of necroptosis with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service