Skip to Content
Merck
All Photos(1)

Key Documents

SML1272

Sigma-Aldrich

I-BET762

≥98% (HPLC)

Synonym(s):

(4S)- 6-(4-Chlorophenyl)-N-ethyl-8-methoxy-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine-4-acetamide, GSK525762, GSK525762A, I-BET

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22ClN5O2
CAS Number:
Molecular Weight:
423.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +80 to +90°, c = 0.3 in methanol

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCNC(C[C@@H]1N=C(C2=CC=C(Cl)C=C2)C3=CC(OC)=CC=C3N4C1=NN=C4C)=O

InChI

1S/C22H22ClN5O2/c1-4-24-20(29)12-18-22-27-26-13(2)28(22)19-10-9-16(30-3)11-17(19)21(25-18)14-5-7-15(23)8-6-14/h5-11,18H,4,12H2,1-3H3,(H,24,29)/t18-/m0/s1

InChI key

AAAQFGUYHFJNHI-SFHVURJKSA-N

Gene Information

Biochem/physiol Actions

I-BET762 (GSK525762) is a selective inhibitor of bromodomain and extra terminal (BET) domain proteins BRD2, BRD3 and BRD4 with IC50 values of 32.5–42.5 nM and no interaction with other bromodomain-containing proteins. I-BET mimicks acetylated histone, preventing the protein-protein interaction between acetyl-lysines and the BET reader protein. This was shown to block expression of inflammatory genes in activated macrophages and confer protection against endotoxic shock and bacterial sepsis. I-BET762 also showed potent anti-myeloma activity in vitro and in vivo.
I-BET762 possesses anti-inflammatory property by controlling the pro-inflammatory gene expression. I-BET762 hinders the MYC (proto-oncogene) expression in cellular models. This action of I-BET762 might serve as an effective therapy in treating prostate cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromodomains: are readers right for epigenetic therapy?.
Conway S J.
ACS Medicinal Chemistry Letters, 3(9), 691?694-691?694 (2012)
Inhibition of BET bromodomain proteins as a therapeutic approach in prostate cancer.
Wyce A, et al.
Oncotarget, 4(12), 2419-2419 (2013)
Francesco Paolo Fiorentino et al.
International journal of molecular sciences, 21(24) (2020-12-20)
Small cell lung cancer (SCLC) is an aggressive type of lung cancer with high mortality that is caused by frequent relapses and acquired resistance. Despite that several target-based approaches with potential therapeutic impact on SCLC have been identified, numerous targeted
Laura Helminen et al.
Nucleic acids research, 52(2), 625-642 (2023-11-28)
Treatment of prostate cancer relies predominantly on the inhibition of androgen receptor (AR) signaling. Despite the initial effectiveness of the antiandrogen therapies, the cancer often develops resistance to the AR blockade. One mechanism of the resistance is glucocorticoid receptor (GR)-mediated
Brian Krug et al.
Cancer cell, 35(5), 782-797 (2019-05-16)
High-grade gliomas defined by histone 3 K27M driver mutations exhibit global loss of H3K27 trimethylation and reciprocal gain of H3K27 acetylation, respectively shaping repressive and active chromatin landscapes. We generated tumor-derived isogenic models bearing this mutation and show that it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service