Skip to Content
Merck
All Photos(1)

Key Documents

SML0492

Sigma-Aldrich

Zanamivir

≥98% (HPLC)

Synonym(s):

4-Guanidino-2,4-dideoxy-2,3-dehydro-N-acetylneuraminic acid, 4-Guanidino-Neu5Ac2en

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N4O7
CAS Number:
Molecular Weight:
332.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +30 to +40°, c = 1 in H2O

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CC(=O)N[C@@H]1[C@@H](NC(N)=N)C=C(O[C@H]1[C@H](O)[C@H](O)CO)C(O)=O

InChI

1S/C12H20N4O7/c1-4(18)15-8-5(16-12(13)14)2-7(11(21)22)23-10(8)9(20)6(19)3-17/h2,5-6,8-10,17,19-20H,3H2,1H3,(H,15,18)(H,21,22)(H4,13,14,16)/t5-,6+,8+,9+,10+/m0/s1

InChI key

ARAIBEBZBOPLMB-UFGQHTETSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Zanamivir has been used:
  • as a neuraminidase inhibitor to study its effects on influenza A virus (IAV) like particles[1]
  • as a neuraminidase inhibitor to study its effects on lipopolysaccharide (LPS)-mediated pathology in sepsis[1]
  • as an antiviral to study its effects on influenza B virus by plaque reduction assay[2]

Biochem/physiol Actions

Zanamivir exhibits therapeutic effects against influenza caused by influenza virus A and B.[3]
Zanamivir is an influenza viral neuraminidase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mila B Ortigoza et al.
mBio, 15(2), e0220323-e0220323 (2024-01-11)
The ongoing transmission of influenza A viruses (IAV) for the past century continues to be a burden to humans. IAV binds terminal sialic acids (SA) of sugar molecules present within the upper respiratory tract (URT) in order to successfully infect
Benjamin O Fulton et al.
Journal of virology, 89(23), 12226-12231 (2015-09-25)
Influenza B virus is a human pathogen responsible for significant health and economic burden. Research into this pathogen has been limited by the lack of reporter viruses. Here we describe the development of both a replication-competent fluorescent influenza B reporter
Synthetic approaches to the neuraminidase inhibitors zanamivir (Relenza) and oseltamivir phosphate (Tamiflu) for the treatment of influenza.
Javier Magano
Chemical reviews, 109(9), 4398-4438 (2009-06-23)
Zanamivir
Waghorn S L and Goa K L
Drugs, 55(5), 721-725 (1998)
Carl J Heneghan et al.
BMJ (Clinical research ed.), 348, g2547-g2547 (2014-05-09)
To describe the potential benefits and harms of zanamivir. Systematic review of clinical study reports of randomised placebo controlled trials and regulatory information Clinical study reports, trial registries, electronic databases, regulatory archives, and correspondence with manufacturers. Randomised placebo controlled trials

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service