Skip to Content
Merck
All Photos(3)

Key Documents

O2378

Sigma-Aldrich

Oxymetazoline hydrochloride

≥99%, solid

Synonym(s):

2-(3-Hydroxy-2,6-dimethyl-4-t-butylbenzyl)-2-imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

form

solid

solubility

H2O: soluble
alcohol: soluble

originator

Schering Plough

SMILES string

Cl.Cc1cc(c(O)c(C)c1CC2=NCCN2)C(C)(C)C

InChI

1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H

InChI key

BEEDODBODQVSIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oxymetazoline hydrochloride is a partial α2A-adrenoceptor agonist; agonist at 5-HT1A, 5-HT1B, and 5-HT1D serotonin receptors. It is a mixed agonist-antagonist at 5-HT1C serotonin receptors.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vittorio Canale et al.
Molecules (Basel, Switzerland), 26(13) (2021-07-03)
The complex pathophysiology of depression, together with the limits of currently available antidepressants, has resulted in the continuous quest for alternative therapeutic strategies. Numerous findings suggest that pharmacological blockade of α2-adrenoceptor might be beneficial for the treatment of depressive symptoms
P Schoeffter et al.
European journal of pharmacology, 196(2), 213-216 (1991-04-17)
Oxymetazoline was recognized with nanomolar affinity by 5-HT1A, 5-HT1B and 5-HT1D binding sites and mimicked the effects of 5-hydroxytryptamine with about the same potency and intrinsic activity as the endogenous amine in the corresponding functional tests. At 5-HT1C receptors, oxymetazoline
H Law et al.
Journal of medicinal chemistry, 41(13), 2243-2251 (1998-06-19)
Benzylimidazolines may represent a class of 5-HT1D ligands that has yet to be exploited. On the basis of a previous report that the 2-(substituted-benzyl)imidazoline alpha-adrenergic agonist oxymetazoline (8) binds with high affinity at calf brain 5-HT1D receptors, we explored the
K Horie et al.
British journal of pharmacology, 116(1), 1611-1618 (1995-09-01)
1. To investigate the structure-activity relationships of alpha-adrenoceptor agonists for the alpha 1-adrenoceptor subtypes, we have compared the imidazoline class of compounds, oxymetazoline and cirazoline, with the phenethylamine, noradrenaline, in their affinities and also in their intrinsic activities in Chinese
Sylwia K Paszun et al.
Acta poloniae pharmaceutica, 70(6), 1079-1085 (2014-01-05)
The presented study aimed at the evaluation of hydrochlorothiazide influence on cilazapril stability in model mixture and fixed dose tablet formulation. The degradation of cilazapril in the presence of hydrochlorothiazide took place according to autocatalytic reaction kinetic mechanism, described mathematically

Articles

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service