Skip to Content
Merck
All Photos(1)

Key Documents

SML0485

Sigma-Aldrich

Azamulin

≥98% (HPLC), powder, pleuromutilin antibiotic

Synonym(s):

(5-Amino-1H-1,2,4-triazol-3-yl)thio]acetic acid (3aS,4R,5S,6R,8R,9R,9aR,10R)-6-ethyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester;, 14-O-(5-(2-Amino1,3,4-triazolyl)thioacetyl)dihydromutilin, Antibiotic TDM 85-530

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38N4O4S
CAS Number:
Molecular Weight:
478.65
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

product name

Azamulin, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC[C@]1(C)C[C@@H](OC(=O)CSc2nnc(N)[nH]2)[C@]3(C)[C@H](C)CC[C@]4(CCC(=O)[C@@H]34)[C@@H](C)[C@@H]1O

InChI

1S/C24H38N4O4S/c1-6-22(4)11-16(32-17(30)12-33-21-26-20(25)27-28-21)23(5)13(2)7-9-24(14(3)19(22)31)10-8-15(29)18(23)24/h13-14,16,18-19,31H,6-12H2,1-5H3,(H3,25,26,27,28)/t13-,14+,16-,18+,19+,22-,23+,24+/m1/s1

InChI key

FMHQJXGMLMSMLC-WBUYAQKGSA-N

General description

Azamulin isolated from fungus Pleurotus mutilis is a diterpene antibiotic. It is related to pleuromutilin being its synthetic azole derivative.

Application

Azamulin has been used as a selective CYP3A4 inhibitor to pretreat primary hepatocyte cells and in prostate cancer cell lines LNCaP and MDAPCa2b.

Biochem/physiol Actions

Azamulin is a derivative of the antibiotic pleuromutilin. The compound is a very specific inhibitor of the CYP3A family (IC50 = 0.03-0.24 μM). Azamulin is 15 and 13 fold more active against CYP3A4 compared to CYP3A5 or CYP3A7, respectively, and is at least 100 fold selective over other CYP isoforms, with the exception of CYP2J2 (approximately 50-fold).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ITag-RNA Allows in Vivo Cell-Type Specific Transcriptional Characterization and Tracking of Circulating Transcripts
Darr J, et al.
Cell, 32(1), 105-112 (2018)
David M Stresser et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(1), 105-112 (2004-01-08)
Azamulin [14-O-(5-(2-amino-1,3,4-triazolyl)thioacetyl)-dihydromutilin] is an azole derivative of the pleuromutilin class of antiinfectives. We tested the inhibition potency of azamulin toward 18 cytochromes P450 using human liver microsomes or microsomes from insect cells expressing single isoforms. In a competitive inhibition model
Priyatham Gorjala et al.
Cancers, 12(4) (2020-04-23)
Androgen receptor signaling is crucial for prostate cancer growth and is positively regulated in part by intratumoral CYP3A5. As African American (AA) men often carry the wild type CYP3A5 and express high levels of CYP3A5 protein, we blocked the wild
Jonatan Darr et al.
Cell reports, 30(9), 3183-3194 (2020-03-05)
Biofluids contain various circulating cell-free RNAs (ccfRNAs). The composition of these ccfRNAs varies among biofluids. They constitute tantalizing biomarker candidates for several pathologies and have been demonstrated to be mediators of cellular communication. Little is known about their function in physiological
Ramakrishna Nirogi et al.
Xenobiotica; the fate of foreign compounds in biological systems, 45(2), 95-106 (2014-07-30)
1. Chemical inhibition is the widely used method in reaction phenotyping assays for estimation of specific enzyme contribution to a given metabolic pathway. The results from phenotyping assays depend on the selectivity of chemical inhibitor and the concentration of inhibitor used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service