Merck
  • iTAG-RNA Isolates Cell-Specific Transcriptional Responses to Environmental Stimuli and Identifies an RNA-Based Endocrine Axis.
iTAG-RNA Isolates Cell-Specific Transcriptional Responses to Environmental Stimuli and Identifies an RNA-Based Endocrine Axis.
Cell reports (2020-03-05)
Jonatan Darr, Archana Tomar, Maximilian Lassi, Raffaele Gerlini, Lucia Berti, Annette Hering, Fabienne Scheid, Martin Hrabě de Angelis, Michael Witting, Raffaele Teperino
ABSTRACT
Biofluids contain various circulating cell-free RNAs (ccfRNAs). The composition of these ccfRNAs varies among biofluids. They constitute tantalizing biomarker candidates for several pathologies and have been demonstrated to be mediators of cellular communication. Little is known about their function in physiological and developmental settings, and most works are limited to in vitro studies. Here, we develop iTAG-RNA, a method for the unbiased tagging of RNA transcripts in mice in vivo. We use iTAG-RNA to isolate hepatocytes and kidney proximal epithelial cell-specific transcriptional responses to a dietary challenge without interfering with the tissue architecture and to identify multiple hepatocyte-secreted ccfRNAs in plasma. We also identify specific transfer of liver-derived ccfRNAs to adipose tissue and skeletal muscle, where they likely constitute a buffering system to maintain lipid homeostasis under acute high-fat-diet feeding. Our findings directly demonstrate in vivo transfer of RNAs between tissues and highlight its implications for endocrine signaling and homeostasis.
Sigma-Aldrich
Azamulin, ≥98% (HPLC)
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
5-Ethynyl uridine, ≥95%
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥2,000 DEA units/mg protein
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
Phosphodiesterase I from Crotalus adamanteus venom, vial of ≥100 units, Purified
Sigma-Aldrich
Adenosine, ≥99%
Millipore
Immobilon®-PSQ PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.2 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.