Skip to Content
Merck
All Photos(1)

Key Documents

SML0285

Sigma-Aldrich

Pleuromutilin

≥95%

Synonym(s):

Drosophilin B, Mutilin 14-glycolate, NSC 121145

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O5
CAS Number:
Molecular Weight:
378.50
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95%

form

powder

optical activity

[α]/D +30 to +40° (c=1; CH2Cl2)

color

white to beige

solubility

DMSO: >10 mg/mL (warmed)

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@@H]1CC[C@@]23CCC(=O)[C@H]2[C@]1(C)[C@@H](C[C@@](C)(C=C)[C@@H](O)[C@@H]3C)OC(=O)CO

InChI

1S/C22H34O5/c1-6-20(4)11-16(27-17(25)12-23)21(5)13(2)7-9-22(14(3)19(20)26)10-8-15(24)18(21)22/h6,13-14,16,18-19,23,26H,1,7-12H2,2-5H3/t13-,14+,16-,18+,19+,20-,21+,22+/m1/s1

InChI key

ZRZNJUXESFHSIO-BKUNHTPHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Pleuromutilin is a diterpene that acts against gram positive bacteria and mycoplasm. It is present in Pleurotus mutilus and Pleurotus passecherianus. Pleuromutilin exhibits its antibacterial action by interfering with the prokaryotic 70S ribosomal subunit and inactivates initiation complex. Pleuromutilin is useful in treating mycoplasma infections in animals.
Pleuromutilin is an antibiotic natural product that inhibits bacterial protein synthesis by binding to bacterial ribosomes in the peptidyl transferase center and inhibiting peptide bond formation.
Pleuromutilin is an antibiotic natural product that inhibits bacterial protein synthesis.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helio S Sader et al.
The Journal of antimicrobial chemotherapy, 67(5), 1170-1175 (2012-01-31)
BC-3781 is an investigational semi-synthetic pleuromutilin antibiotic, which recently finished a clinical Phase 2 trial in acute bacterial skin and skin structure infections. BC-3781 binds to the 50S ribosomal subunit and cross-resistance with other antimicrobial classes is uncommon. We evaluated
Lykke H Hansen et al.
Antimicrobial agents and chemotherapy, 56(7), 3563-3567 (2012-05-02)
The cfr gene encodes the Cfr methyltransferase that methylates a single adenine in the peptidyl transferase region of bacterial ribosomes. The methylation provides resistance to several classes of antibiotics that include drugs of clinical and veterinary importance. This paper describes
Xinyang Wang et al.
Bioorganic & medicinal chemistry letters, 22(19), 6166-6172 (2012-08-31)
Owing to the increasingly serious problems caused by multidrug resistance in community-acquired infection pathogens, it has become an urgent need to develop new classes of antibiotics for overcoming the resistance. In this paper, we describe the design and synthesis of
Encyclopedia of Food Sciences and Nutrition (Second Edition), 575-575 (2014)
Thomas J K Findley et al.
Organic & biomolecular chemistry, 9(7), 2433-2451 (2011-02-18)
The cis-hydrindane motif is found in a number of natural products that display important biological activity. A flexible, stereoselective approach to the framework has been developed that features highly diastereoselective, SmI(2)-mediated cyclisations. The strategy has been exploited in the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service