Skip to Content
Merck
All Photos(1)

Key Documents

SML0284

Sigma-Aldrich

Mycophenolate mofetil

≥98% (HPLC)

Synonym(s):

(4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid 2-(4-morpholinyl)ethyl ester, RS 61443, TM-MMF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31NO7
CAS Number:
Molecular Weight:
433.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥15 mg/mL

storage temp.

room temp

SMILES string

COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)CCC(=O)OCCN3CCOCC3

InChI

1S/C23H31NO7/c1-15(5-7-19(25)30-13-10-24-8-11-29-12-9-24)4-6-17-21(26)20-18(14-31-23(20)27)16(2)22(17)28-3/h4,26H,5-14H2,1-3H3/b15-4+

InChI key

RTGDFNSFWBGLEC-SYZQJQIISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Mycophenolate mofetil has been used to treat wild-type embryos for inhibiting nucleotide synthesis.

Biochem/physiol Actions

Mycophenolate mofetil functions as a rate-limiting enzyme in de novo synthesis of guanosine nucleotides.
Mycophenolate mofetil is a prodrug of mycophenolic acid (Cat. # M5255) that is cleaved by nonspecific esterases in vivo to produce the parent compound. Mycophenolic acid blocks inosine monophosphate dehydrogenase and is a potent immunosuppresive agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mycophenolate mofetil and its mechanisms of action
Allison AC and Eugui EM
Immunopharmacology, 47(2-3), 85-118 (2000)
Wuhong Pei et al.
Scientific reports, 6, 29946-29946 (2016-07-19)
Phosphoribosyl pyrophosphate synthetase-1 (PRPS1) is a key enzyme in nucleotide biosynthesis, and mutations in PRPS1 are found in several human diseases including nonsyndromic sensorineural deafness, Charcot-Marie-Tooth disease-5, and Arts Syndrome. We utilized zebrafish as a model to confirm that mutations
Eduardo F Mysler et al.
Arthritis and rheumatism, 65(9), 2368-2379 (2013-06-07)
To investigate the efficacy and safety of ocrelizumab in patients with class III/IV lupus nephritis (LN). Patients were randomized 1:1:1 to receive placebo, 400 mg ocrelizumab, or 1,000 mg ocrelizumab given as an intravenous infusion on days 1 and 15
Christopher J Yates et al.
Transplantation, 96(8), 726-731 (2013-08-02)
The sensitivity of fasting plasma glucose (FPG) in screening for new-onset diabetes after transplantation (NODAT) has been questioned, particularly in the presence of moderate-dose prednisolone, where peak plasma glucose occurs 7 to 8 hr after administration. Oral glucose tolerance testing
K Budde et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 14(12), 2796-2806 (2014-10-04)
This Phase III randomized trial examined efficacy and safety of a novel once-daily extended-release tacrolimus formulation (LCP-Tacro [LCPT]) versus twice-daily tacrolimus in de novo kidney transplantation. Primary efficacy end point was proportion of patients with treatment failure (death, graft failure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service