Merck
  • Novel once-daily extended-release tacrolimus (LCPT) versus twice-daily tacrolimus in de novo kidney transplants: one-year results of Phase III, double-blind, randomized trial.
Novel once-daily extended-release tacrolimus (LCPT) versus twice-daily tacrolimus in de novo kidney transplants: one-year results of Phase III, double-blind, randomized trial.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2014-10-04)
K Budde, S Bunnapradist, J M Grinyo, K Ciechanowski, J E Denny, H T Silva, L Rostaing
ABSTRACT
This Phase III randomized trial examined efficacy and safety of a novel once-daily extended-release tacrolimus formulation (LCP-Tacro [LCPT]) versus twice-daily tacrolimus in de novo kidney transplantation. Primary efficacy end point was proportion of patients with treatment failure (death, graft failure, biopsy-proven acute rejection or lost to follow-up) within 12 months. Starting doses were, LCPT: 0.17 mg/kg/day and tacrolimus twice-daily: 0.1 mg/kg/day; 543 patients were randomized, LCPT: n = 268; tacrolimus twice-daily: n = 275. At 12 months treatment failure was LCPT: 18.3% and tacrolimus twice-daily: 19.6%; the upper 95% CI of the treatment difference was +5.27%, below the predefined +10% noninferiority criteria. There were no significant differences in the incidence of individual efficacy events or adverse events. Target tacrolimus trough levels were more rapidly achieved in the LCPT group. Following initial dose, 36.6% of patients in the LCPT group had rapidly attained trough levels within 6-11 ng/mL versus 18.5% of tacrolimus twice-daily patients; majority of tacrolimus twice-daily patients (74.7%) had troughs <6 ng/mL compared with 33.5% in the LCPT group. Overall, cumulative study dose was 14% lower for LCPT. Results suggest that use of once-daily LCPT in de novo kidney transplantation is efficacious and safe. Lower LCPT dose reflects the improved absorption provided by the novel formulation.
Product Description
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
USP
Mycophenolate mofetil, United States Pharmacopeia (USP) Reference Standard
Supelco
Mycophenolate Mofetil, Pharmaceutical Secondary Standard; Certified Reference Material