Skip to Content
Merck
All Photos(1)

Key Documents

SML0256

Sigma-Aldrich

2OHOA

≥98% (HPLC)

Synonym(s):

(9Z)-2-Hydroxy-9-octadecenoic acid; cis-2-Hydroxy-9-octadecenoic acid, 2-Hydroxy Oleic Acid, Minerval

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O3
CAS Number:
Molecular Weight:
298.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

storage condition

protect from light

color

white to tan

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

CCCCCCCC\C=C/CCCCCCC(O)C(O)=O

InChI

1S/C18H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(19)18(20)21/h9-10,17,19H,2-8,11-16H2,1H3,(H,20,21)/b10-9-

InChI key

JBSOOFITVPOOSY-KTKRTIGZSA-N

Application

2OHOA has been used as:
  • a modulator of the sphingolipid and glycerophospholipid pathway to study its effects on myelination in mice Charcot–Marie–Tooth type 1A (CMT1A) myelinopathy and myelinating dorsal root ganglia cultures
  • in biophysical studies to study its effects on artificial membranes
  • to study its interactions with major membrane lipid films

Biochem/physiol Actions

2-Hydroxy oleic acid (2OHOA) is a derivative of oleic acid. It plays a role in membrane organization, alterations in membrane fluidity, an increase of the tendency to form non-lamellar structures, and remodeling of microdomains by interacting with the lipids in membranes of pathological cells.
2OHOA induces cell cycle arrest and apoptosis in several cancer cell lines, including glioma, leukemia, breast and colon cancer lines. 2OHOA increases sphingomyelin (SM) levels in the membranes of tumor cells, which typically display decreased SM membrane content, and remodeled membranes, compared with normal cells. The compound has no effect on SM levels in non-cancer cells.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karolina Węder et al.
Biochimica et biophysica acta. Biomembranes, 1860(11), 2329-2336 (2018-06-05)
2-Hydroxyoleic acid (2OHOA, Minerval), a derivative of oleic acid, is the lipid used in Membrane Lipid Therapy. This compound is of confirmed anticancer effect, however its exact mechanism of action has not been fully elucidated. In this work the interactions
The influence of 2-hydroxyoleic acid--an anticancer drug--on model membranes of different fluidity modulated by the cholesterol content
Olechowska K, et al.
Journal of Molecular Liquids, 283, 756-762 (2019)
Maria Laura Martin et al.
Journal of lipid research, 54(5), 1457-1465 (2013-03-09)
The mechanism of action of 2-hydroxyoleic acid (2OHOA), a potent antitumor drug, involves the rapid and specific activation of sphingomyelin synthase (SMS), leading to a 4-fold increase in SM mass in tumor cells. In the present study, we investigated the
Amaia Marcilla-Etxenike et al.
PloS one, 7(10), e48235-e48235 (2012-11-08)
2-Hydroxyoleic acid is a synthetic fatty acid with potent anti-cancer activity which does not induce undesired side effects. However, the molecular and cellular mechanisms by which this compound selectively kills human glioma cancer cells without killing normal cells is not
Nuanyi Liang et al.
Journal of agricultural and food chemistry, 68(32), 8648-8657 (2020-07-17)
The quality of fermented sausage is strongly influenced by its fatty acid (FA). However, the role of a defined starter culture in modifying sausage FA composition, and especially in the production of hydroxy FAs (HFAs), has not been determined. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service