Skip to Content
Merck
All Photos(2)

Key Documents

219800

Sigma-Aldrich

2-Hydroxyhexanoic acid

98%

Synonym(s):

DL-α-Hydroxycaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CO2H
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

60-62 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

CCCCC(O)C(O)=O

InChI

1S/C6H12O3/c1-2-3-4-5(7)6(8)9/h5,7H,2-4H2,1H3,(H,8,9)

InChI key

NYHNVHGFPZAZGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Solubility of 2-hydroxyhexanoic acid in supercritical CO2 has been studied.

Application

2-Hydroxyhexanoic acid was used in the synthesis of aliphatic copolyesters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Liu et al.
Environmental technology, 38(21), 2639-2649 (2016-12-15)
Carbon substrate is required by biological nutrient removal (BNR) microorganism, but it is usually insufficient in the influent of many municipal wastewater treatment plants. In this study the use of ethanol-enriched fermentation liquid, which was derived from dairy wastewater, as
Solubilities of lactic acid and 2-hydroxyhexanoic acid in supercritical CO2.
Gregorowicz J.
Fluid Phase Equilibria, 166(1), 39-46 (1999)
L I Woolf et al.
Journal of chromatography, 231(2), 237-245 (1982-09-10)
Plasma or whole blood is treated with o-phenylenediamine dihydrochloride in phosphoric acid under conditions found spectrophotometrically to give maximum yields of the quinoxalinols. The quinoxalinols are extracted and, after removing phosphoric acid, etc., silylated with bis-trimethylsilyltrifluoracetamide in acetonitrile. Other solvents
J B Brooks et al.
Journal of clinical microbiology, 25(2), 445-448 (1987-02-01)
Serum (SR) and cerebrospinal fluid (CSF) from a patient suspected of having tuberculous meningitis were submitted to our laboratory for analysis by frequency-pulsed electron capture gas-liquid chromatography (FPEC GLC). The samples were tested for the presence of carboxylic acids, alcohols
Grazyna Adamus et al.
Rapid communications in mass spectrometry : RCM, 18(13), 1436-1446 (2004-06-25)
A series of aliphatic copolyesters was obtained from (R,S)-beta-butyrolactone and two isomeric hydroxy acids, 6-hydroxyhexanoic and (R,S)-2-hydroxyhexanoic acids. The reactions were conducted in bulk without catalyst. Electrospray ionization tandem mass spectrometry (ESI-MSn) was used for molecular characterization of these copolyester

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service